Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          Assay Name: hsa-miR-193a-5p
          miRBase Accession Number: MI0000487
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 31559996 - 31560083 [+] on Build GRCh38
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Species: Human, Bovine, Chicken, Dasypus novemcinctus, Dog, Guinea pig, Horse, Ophiophagus hannah, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Taeniopygia guttata
          Product Type: QuantiGene Singleplex miRNA
          Assay ID SM-10112
          Size 200 rxns
          Availability Made To Order
          Catalog # QGSM-200
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000082, mir-193

          Mature miRNA Sequence:

          UGGGUCUUUGCGGGCGAGAUGA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-193a-5p MIMAT0004614
          Bovine bta-miR-193a-5p MIMAT0003794
          Chicken gga-miR-193a-5p MIMAT0026787
          Dasypus novemcinctus dno-miR-193a-5p MIMAT0047721
          Dog cfa-miR-193a MIMAT0006735
          Guinea pig cpo-miR-193a-5p MIMAT0047070
          View More Rows
          Horse eca-miR-193a-5p MIMAT0013025
          Ophiophagus hannah oha-miR-193-5p MIMAT0036779
          Pig ssc-miR-193a-5p MIMAT0013894
          Pongo pygmaeus ppy-miR-193a-5p MIMAT0015787
          Pteropus alecto pal-miR-193a-5p MIMAT0039931
          Python bivittatus pbv-miR-193a-5p MIMAT0038989
          Rabbit ocu-miR-193a-5p MIMAT0048303
          Rhesus monkey mml-miR-193a-5p MIMAT0006225
          Taeniopygia guttata tgu-miR-193b-5p MIMAT0014654

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-193a
          Stem-loop Accession # MI0000487
          Stem-loop Sequence
          CGAGGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUUCUCGGCCCCCG
          Chromosome Location Chr. 17 - 31559996 - 31560083 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004614
          miRBase ID: hsa-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: Chr. 17 - 31559996 - 31560083 [+] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-193a
          Stem-loop Accession # MI0005014
          Stem-loop Sequence
          UGGGAGCUGAGAGCUGGGUCUUUGCGGGCGAGAUGAAGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003794
          miRBase ID: bta-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-193a
          Stem-loop Accession # MI0007564
          Stem-loop Sequence
          AGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGAGGUUCGUGUCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026787
          miRBase ID: gga-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-193a
          Stem-loop Accession # MI0039024
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047721
          miRBase ID: dno-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-193a
          Stem-loop Accession # MI0008159
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006735
          miRBase ID: cfa-miR-193a
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-193a
          Stem-loop Accession # MI0038681
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047070
          miRBase ID: cpo-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-193a
          Stem-loop Accession # MI0012773
          Stem-loop Sequence
          GGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCAGUUCAACUGGCCUACAAAGUCCCAGUCUUCGGCCCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013025
          miRBase ID: eca-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-193
          Stem-loop Accession # MI0031387
          Stem-loop Sequence
          UGGGGCUGGGUCUUUGCGGGCGAGAUGAGAAGUUUCUGCCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036779
          miRBase ID: oha-miR-193-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-193a
          Stem-loop Accession # MI0013113
          Stem-loop Sequence
          GCGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUUGGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013894
          miRBase ID: ssc-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-193a
          Stem-loop Accession # MI0014855
          Stem-loop Sequence
          CGAGGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAAGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015787
          miRBase ID: ppy-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-193a
          Stem-loop Accession # MI0032406
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGAUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039931
          miRBase ID: pal-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-193a
          Stem-loop Accession # MI0030265
          Stem-loop Sequence
          GAGCAGCUGGGAGUUUGGGGCUGGGUCUUUGCGGGCGAGAUGAGAAGUUUCUGCCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCUCUUAACUUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038989
          miRBase ID: pbv-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-193a
          Stem-loop Accession # MI0039330
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048303
          miRBase ID: ocu-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-193a
          Stem-loop Accession # MI0007655
          Stem-loop Sequence
          GGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006225
          miRBase ID: mml-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-193b
          Stem-loop Accession # MI0013838
          Stem-loop Sequence
          GCUGGGUCUUUGCGGGCGAGAUGAGAGGUUCCUGUCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014654
          miRBase ID: tgu-miR-193b-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          QuantiGene Singleplex miRNA Control : SMC-10112


          miRBase Alias:

          Human: hsa-miR-193a-5p (v22)
          Bos taurus: bta-miR-193a-5p (v22)
          Citrobacter farmeri: cfa-miR-193a (v22)
          Citrobacter portucalensis: cpo-miR-193a-5p (v22)
          Dasypus novemcinctus: dno-miR-193a-5p (v22)
          Enterovirus CA55-1988: eca-miR-193a-5p (v22)
          Gallus gallus: gga-miR-193a-5p (v22)
          Macaca mulatta: mml-miR-193a-5p (v22)
          Ophiophagus hannah: oha-miR-193-5p (v22)
          Rabbit: ocu-miR-193a-5p (v22)
          Pongo pygmaeus: ppy-miR-193a-5p (v22)
          Providencia alcalifaciens: pal-miR-193a-5p (v22)
          Python bivittatus: pbv-miR-193a-5p (v22)
          Staphylococcus schleiferi: ssc-miR-193a-5p (v22)
          Trabulsiella guamensis ATCC 49490: tgu-miR-193b-5p (v22)

          Back To Top

          Related Products

          • QuantiGene Singleplex Assay Kit
          • QuantiGene Singleplex HT Assay Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline