Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          Assay Name: hsa-miR-193a-5p
          miRBase Accession Number: MI0000487
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 31559996 - 31560083 [+] on Build GRCh38
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Species: Human, Bovine, Chicken, Dasypus novemcinctus, Dog, Guinea pig, Horse, Ophiophagus hannah, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Taeniopygia guttata
          Product Type: QuantiGene Singleplex miRNA Control
          Assay ID SMC-10112
          Size 30 std curves
          Availability Made To Order
          Catalog # QGSMC
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000082, mir-193

          Mature miRNA Sequence:

          UGGGUCUUUGCGGGCGAGAUGA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-193a-5p MIMAT0004614
          Bovine bta-miR-193a-5p MIMAT0003794
          Chicken gga-miR-193a-5p MIMAT0026787
          Dasypus novemcinctus dno-miR-193a-5p MIMAT0047721
          Dog cfa-miR-193a MIMAT0006735
          Guinea pig cpo-miR-193a-5p MIMAT0047070
          View More Rows
          Horse eca-miR-193a-5p MIMAT0013025
          Ophiophagus hannah oha-miR-193-5p MIMAT0036779
          Pig ssc-miR-193a-5p MIMAT0013894
          Pongo pygmaeus ppy-miR-193a-5p MIMAT0015787
          Pteropus alecto pal-miR-193a-5p MIMAT0039931
          Python bivittatus pbv-miR-193a-5p MIMAT0038989
          Rabbit ocu-miR-193a-5p MIMAT0048303
          Rhesus monkey mml-miR-193a-5p MIMAT0006225
          Taeniopygia guttata tgu-miR-193b-5p MIMAT0014654

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-193a
          Stem-loop Accession # MI0000487
          Stem-loop Sequence
          CGAGGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUUCUCGGCCCCCG
          Chromosome Location Chr. 17 - 31559996 - 31560083 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004614
          miRBase ID: hsa-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: Chr. 17 - 31559996 - 31560083 [+] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-193a
          Stem-loop Accession # MI0005014
          Stem-loop Sequence
          UGGGAGCUGAGAGCUGGGUCUUUGCGGGCGAGAUGAAGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003794
          miRBase ID: bta-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-193a
          Stem-loop Accession # MI0007564
          Stem-loop Sequence
          AGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGAGGUUCGUGUCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026787
          miRBase ID: gga-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-193a
          Stem-loop Accession # MI0039024
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047721
          miRBase ID: dno-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-193a
          Stem-loop Accession # MI0008159
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006735
          miRBase ID: cfa-miR-193a
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-193a
          Stem-loop Accession # MI0038681
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047070
          miRBase ID: cpo-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-193a
          Stem-loop Accession # MI0012773
          Stem-loop Sequence
          GGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCAGUUCAACUGGCCUACAAAGUCCCAGUCUUCGGCCCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013025
          miRBase ID: eca-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-193
          Stem-loop Accession # MI0031387
          Stem-loop Sequence
          UGGGGCUGGGUCUUUGCGGGCGAGAUGAGAAGUUUCUGCCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036779
          miRBase ID: oha-miR-193-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-193a
          Stem-loop Accession # MI0013113
          Stem-loop Sequence
          GCGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUUGGUUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013894
          miRBase ID: ssc-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-193a
          Stem-loop Accession # MI0014855
          Stem-loop Sequence
          CGAGGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAAGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015787
          miRBase ID: ppy-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-193a
          Stem-loop Accession # MI0032406
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGAUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039931
          miRBase ID: pal-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-193a
          Stem-loop Accession # MI0030265
          Stem-loop Sequence
          GAGCAGCUGGGAGUUUGGGGCUGGGUCUUUGCGGGCGAGAUGAGAAGUUUCUGCCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCUCUUAACUUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038989
          miRBase ID: pbv-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-193a
          Stem-loop Accession # MI0039330
          Stem-loop Sequence
          UGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGUUCAACUGGCCUACAAAGUCCCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048303
          miRBase ID: ocu-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-193a
          Stem-loop Accession # MI0007655
          Stem-loop Sequence
          GGAUGGGAGCUGAGGGCUGGGUCUUUGCGGGCGAGAUGAGGGUGUCGGAUCAACUGGCCUACAAAGUCCCAGUCCUCGGCCCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006225
          miRBase ID: mml-miR-193a-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-193b
          Stem-loop Accession # MI0013838
          Stem-loop Sequence
          GCUGGGUCUUUGCGGGCGAGAUGAGAGGUUCCUGUCUUCAACUGGCCUACAAAGUCCCAGUUCUCGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014654
          miRBase ID: tgu-miR-193b-5p
          Mature miRNA Sequence: UGGGUCUUUGCGGGCGAGAUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          QuantiGene Singleplex miRNA : SM-10112


          miRBase Alias:

          Human: hsa-miR-193a-5p (v22)
          Bos taurus: bta-miR-193a-5p (v22)
          Citrobacter farmeri: cfa-miR-193a (v22)
          Citrobacter portucalensis: cpo-miR-193a-5p (v22)
          Dasypus novemcinctus: dno-miR-193a-5p (v22)
          Enterovirus CA55-1988: eca-miR-193a-5p (v22)
          Gallus gallus: gga-miR-193a-5p (v22)
          Macaca mulatta: mml-miR-193a-5p (v22)
          Ophiophagus hannah: oha-miR-193-5p (v22)
          Rabbit: ocu-miR-193a-5p (v22)
          Pongo pygmaeus: ppy-miR-193a-5p (v22)
          Providencia alcalifaciens: pal-miR-193a-5p (v22)
          Python bivittatus: pbv-miR-193a-5p (v22)
          Staphylococcus schleiferi: ssc-miR-193a-5p (v22)
          Trabulsiella guamensis ATCC 49490: tgu-miR-193b-5p (v22)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline