Search
Search

| Assay Name: | hsa-miR-142-3p |
|---|---|
| miRBase Accession Number: | MI0000458 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUAGUGUUUCCUACUUUAUGGA |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Artibeus jamaicensis, Eptesicus fuscus, Horse, Monodelphis domestica, Ophiophagus hannah, Pan troglodytes, Pongo pygmaeus, Rhesus monkey, Xenopus laevis, Xenopus tropicalis, Zebrafish |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000084, mir-142 |
Mature miRNA Sequence: | UGUAGUGUUUCCUACUUUAUGGA |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-142-3p | MIMAT0000434 | |
| Mouse | mmu-miR-142a-3p | MIMAT0000155 | |
| Rat | rno-miR-142-3p | MIMAT0000848 | |
| Alligator mississippiensis | ami-miR-142-3p | MIMAT0038163 | |
| Artibeus jamaicensis | aja-miR-142 | MIMAT0031136 | |
| Eptesicus fuscus | efu-miR-142 | MIMAT0035200 | |
| View More Rows | |||
| Horse | eca-miR-142-3p | MIMAT0013023 | |
| Monodelphis domestica | mdo-miR-142 | MIMAT0004112 | |
| Ophiophagus hannah | oha-miR-142-3p | MIMAT0036722 | |
| Pan troglodytes | ptr-miR-142 | MIMAT0008037 | |
| Pongo pygmaeus | ppy-miR-142-3p | MIMAT0015766 | |
| Rhesus monkey | mml-miR-142-3p | MIMAT0006200 | |
| Xenopus laevis | xla-miR-142-3p | MIMAT0046455 | |
| Xenopus tropicalis | xtr-miR-142-3p | MIMAT0003603 | |
| Zebrafish | dre-miR-142a-3p | MIMAT0003160 | |