Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002326
          Assay Name: hsa-miR-216b
          miRBase Accession Number: MI0005569
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 56000714 - 56000795 [-] on Build GRCh38
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Species: Human, Mouse, Rat, Anolis carolinensis, Bovine, Callithrix jacchus, Chicken, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Dog, Guinea pig, Horse, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002326
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000054, mir-216

          Mature miRNA Sequence:

          AAAUCUCUGCAGGCAAAUGUGA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-216b-5p MIMAT0004959
          Mouse mmu-miR-216b-5p MIMAT0003729
          Rat rno-miR-216b-5p MIMAT0017846
          Anolis carolinensis aca-miR-216b-5p MIMAT0021876
          Bovine bta-miR-216b MIMAT0009266
          Callithrix jacchus cja-miR-216b MIMAT0039342
          View More Rows
          Chicken gga-miR-216b MIMAT0007748
          Columba livia cli-miR-216b-5p MIMAT0038598
          Cricetulus griseus cgr-miR-216b MIMAT0041157
          Dasypus novemcinctus dno-miR-216b-5p MIMAT0047764
          Dog cfa-miR-216b MIMAT0006592
          Guinea pig cpo-miR-216b-5p MIMAT0047121
          Horse eca-miR-216b MIMAT0013073
          Pan troglodytes ptr-miR-216b MIMAT0008072
          Pongo pygmaeus ppy-miR-216b MIMAT0015802
          Pteropus alecto pal-miR-216b-5p MIMAT0040051
          Python bivittatus pbv-miR-216b-5p MIMAT0039024
          Rhesus monkey mml-miR-216b MIMAT0006243

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-216b
          Stem-loop Accession # MI0005569
          Stem-loop Sequence
          GCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAGGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGACA
          Chromosome Location Chr. 2 - 56000714 - 56000795 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004959
          miRBase ID: hsa-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: Chr. 2 - 56000714 - 56000795 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-216b
          Stem-loop Accession # MI0004126
          Stem-loop Sequence
          UUGGCAGACUGGGAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAACCACACACUUACCUGUAGAGAUUCUUCAGUCUGACAA
          Chromosome Location Chr. 11 - 28746191 - 28746276 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003729
          miRBase ID: mmu-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: Chr. 11 - 28746191 - 28746276 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-216b
          Stem-loop Accession # MI0015435
          Stem-loop Sequence
          UGCACGUUGGCAGACUGGGAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAAUCACACACUUACUUGUAGAGAUUCUUCAGUCUGACAACAUCAAGCAU
          Chromosome Location Chr. 14 - 113101078 - 113101179 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017846
          miRBase ID: rno-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: Chr. 14 - 113101078 - 113101179 [+] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-216b
          Stem-loop Accession # MI0018820
          Stem-loop Sequence
          UCAAAGUCACAGACUGAGAAAUCUCUGCAGGCAAAUGUGAUGUCUUGACCAGCAAUCUCACAGUUACCAGCAGAGGUUCUGCAAUCUGGACUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021876
          miRBase ID: aca-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-216b
          Stem-loop Accession # MI0009778
          Stem-loop Sequence
          GCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUAAAGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009266
          miRBase ID: bta-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-216b
          Stem-loop Accession # MI0031855
          Stem-loop Sequence
          CUUCAAAGUGGCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAAUCACACAGUUACCCGCAGAGAUUCUACAGUCUGACACCUUCAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039342
          miRBase ID: cja-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-216b
          Stem-loop Accession # MI0008205
          Stem-loop Sequence
          GAGUCACAGACUGGGAAAUCUCUGCAGGCAAAUGUGAUGUCUUUAUAGCAAUCUCACAAUUACCUGUAGAGAUUCUACAAUCUGGCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007748
          miRBase ID: gga-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-216b
          Stem-loop Accession # MI0030018
          Stem-loop Sequence
          GUCGCAGACUGGGAAAUCUCUGCAGGCAAAUGUGAUGUCUUUAUAGCAAUCUCACAAUUACCUGUAGAGAUUCUACAAUCUGGCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038598
          miRBase ID: cli-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-216b
          Stem-loop Accession # MI0033086
          Stem-loop Sequence
          GUGGCAGACUGAGAAAUCUCUGCAGGCAAAUGUGAUGUCAGUGAAGAAAUCACACACUUACCUGUAGAGAUUCUUCAGUCUGUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041157
          miRBase ID: cgr-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-216b
          Stem-loop Accession # MI0039046
          Stem-loop Sequence
          AAAUCUCUGCAGGCAAAUGUGAUGUCACUAAAGAAAUCACACACUUACCUAUAGAGAUUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047764
          miRBase ID: dno-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-216b
          Stem-loop Accession # MI0007987
          Stem-loop Sequence
          AAAUCUCUGCAGGCAAAUGUGAUGUCACUAAAGAAAUCACACACUUACCCGUAGAGAUUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006592
          miRBase ID: cfa-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-216b
          Stem-loop Accession # MI0038708
          Stem-loop Sequence
          AAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAUCACACACUUACCUGUAAAGAUUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047121
          miRBase ID: cpo-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-216b
          Stem-loop Accession # MI0012818
          Stem-loop Sequence
          GUGGCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUAAAGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGACAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013073
          miRBase ID: eca-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-216b
          Stem-loop Accession # MI0008582
          Stem-loop Sequence
          GCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAGGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008072
          miRBase ID: ptr-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-216b
          Stem-loop Accession # MI0014867
          Stem-loop Sequence
          GCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015802
          miRBase ID: ppy-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-216b
          Stem-loop Accession # MI0032468
          Stem-loop Sequence
          AAAUCUCUGCAGGCAAAUGUGAUGUCACUAAAGAAAUCACACACUUACCUGUAGAGAUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040051
          miRBase ID: pal-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-216b
          Stem-loop Accession # MI0030290
          Stem-loop Sequence
          GGUCCCAGUCACAGAUUGAGAAAUCUCUGCAGGCAAAUGUGAUGUCAUGAUUGCAAUCUCACAGUUACCAGUAGAGGUUCUGCAAUCUGGAGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039024
          miRBase ID: pbv-miR-216b-5p
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-216b
          Stem-loop Accession # MI0007673
          Stem-loop Sequence
          GCAGACUGGAAAAUCUCUGCAGGCAAAUGUGAUGUCACUGAAGAAAUCACACACUUACCCGUAGAGAUUCUACAGUCUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006243
          miRBase ID: mml-miR-216b
          Mature miRNA Sequence: AAAUCUCUGCAGGCAAAUGUGA
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool A
          Megaplex™ RT Primers, Human Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307782_pri
          TaqMan™ Pri-miRNA Assay : Rn04229467_pri
          TaqMan™ Pri-miRNA Assay : Hs03305200_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12302
          Ambion® Pre-miR™ miRNA Precursor : PM12302
          mirVana® miRNA inhibitor : MH12302
          mirVana® miRNA mimic : MC12302
          TaqMan™ Advanced miRNA Assay : mmu479456_mir
          TaqMan™ Advanced miRNA Assay : 479456_mir
          TaqMan™ Advanced miRNA Assay : rno479456_mir
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          FGMIR : TaqManTM Rodent MicroRNA Array A
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          FGMIR : TaqManTM Human MicroRNA Array A
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0


          miRBase Alias:

          Human: hsa-miR-216b (v19)
          Mouse: mmu-miR-216b (v17)
          Aeromonas caviae Ae398: aca-miR-216b (v18)
          Citrobacter farmeri: cfa-miR-216 (v12)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline