Search
Search

| Assay Name: | tgu-miR-155* |
|---|---|
| miRBase Accession Number: | MI0013842 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCCUACACGUUAGCAUUAACA |
| Species: | Taeniopygia guttata |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000157, mir-155 |
Mature miRNA Sequence: | CUCCUACACGUUAGCAUUAACA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Taeniopygia guttata | tgu-miR-155-3p | MIMAT0014656 |