Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 462049_mat
          Assay Name: mmu-miR-1a-1*
          miRBase Accession Number: MI0000139
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 180389048 - 180389124 [+] on Build GRCm38
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Species: Mouse, Chicken, Chrysemys picta, Columba livia, Dasypus novemcinctus, Ictalurus punctatus, Ophiophagus hannah, Paralichthys olivaceus, Rhesus monkey, Salmo salar, Taeniopygia guttata
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 462049_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000038, mir-1

          Mature miRNA Sequence:

          ACAUACUUCUUUAUAUGCCCAUA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-1a-1-5p MIMAT0016979
          Chicken gga-miR-1a-1-5p MIMAT0031071
          Chrysemys picta cpi-miR-1a-5p MIMAT0037612
          Columba livia cli-miR-1a-5p MIMAT0038391
          Dasypus novemcinctus dno-miR-1-5p MIMAT0047488
          Ictalurus punctatus ipu-miR-1 MIMAT0029384
          View More Rows
          Ophiophagus hannah oha-miR-1b-5p MIMAT0036799
          Paralichthys olivaceus pol-miR-1-5p MIMAT0025412
          Rhesus monkey mml-miR-1-1-5p MIMAT0026788
          Salmo salar ssa-miR-1-4-5p MIMAT0032474
          Taeniopygia guttata tgu-miR-1-1-5p MIMAT0031087

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-1a-1
          Stem-loop Accession # MI0000139
          Stem-loop Sequence
          GCUUGGGACACAUACUUCUUUAUAUGCCCAUAUGAACCUGCUAAGCUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGC
          Chromosome Location Chr. 2 - 180389048 - 180389124 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0016979
          miRBase ID: mmu-miR-1a-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: Chr. 2 - 180389048 - 180389124 [+] on Build GRCm38

          Chicken

          Stem-loop ID gga-mir-1a-1
          Stem-loop Accession # MI0001247
          Stem-loop Sequence
          UGAGAGACAUACUUCUUUAUAUGCCCAUAUGAACCUGGCAAUCUAUGGAAUGUAAAGAAGUAUGUAUUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031071
          miRBase ID: gga-miR-1a-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-1a-1
          Stem-loop Accession # MI0029384
          Stem-loop Sequence
          CUACCUGCUUGAGAGACAUACUUCUUUAUAUGCCCAUAUGAACCUGGCAAGCUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGUGGGGAUCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037612
          miRBase ID: cpi-miR-1a-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-1a-1
          Stem-loop Accession # MI0029893
          Stem-loop Sequence
          CUGCUUGAGAGACAUACUUCUUUAUAUGCCCAUAUGAACCUGGGAAGCUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038391
          miRBase ID: cli-miR-1a-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-1-1
          Stem-loop Accession # MI0038895
          Stem-loop Sequence
          ACAUACUUCUUUAUAUGCCCAUAUGGAGCUCGGAGCUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047488
          miRBase ID: dno-miR-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-1-1
          Stem-loop Accession # MI0024455
          Stem-loop Sequence
          ACAUACUUCUUUAUAUGCCCAUAUGAACAAGAGCAGCUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029384
          miRBase ID: ipu-miR-1
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-1b
          Stem-loop Accession # MI0031397
          Stem-loop Sequence
          AAUGACAUACUUCUUUAUAUGCCCAUAUGAACCUGGCCAACUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGCACGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036799
          miRBase ID: oha-miR-1b-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Paralichthys olivaceus

          Stem-loop ID pol-mir-1
          Stem-loop Accession # MI0022189
          Stem-loop Sequence
          ACAUACUUCUUUAUAUGCCCAUAUGAACAAGAGCAACUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025412
          miRBase ID: pol-miR-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-1-1
          Stem-loop Accession # MI0007568
          Stem-loop Sequence
          GCUUGGGAAACAUACUUCUUUAUAUGCCCAUAUGGACCUGCUAAGCUAUGGAAUGUAAAGAAGUAUGUAUCUCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026788
          miRBase ID: mml-miR-1-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-1-4
          Stem-loop Accession # MI0026580
          Stem-loop Sequence
          ACAUACUUCUUUAUAUGCCCAUAUGAACAAGAGCAACUAUGGAAUGUAAAGAAGUAUGUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032474
          miRBase ID: ssa-miR-1-4-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-1-1
          Stem-loop Accession # MI0013781
          Stem-loop Sequence
          GACAUACUUCUUUAUAUGCCCAUAUGAACCUGGCAAGCUAUGGAAUGUAAAGAAGUAUGUAUUUCAGGUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031087
          miRBase ID: tgu-miR-1-1-5p
          Mature miRNA Sequence: ACAUACUUCUUUAUAUGCCCAUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306163_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19454
          Ambion® Pre-miR™ miRNA Precursor : PM19454
          mirVana® miRNA inhibitor : MH19454
          mirVana® miRNA mimic : MC19454


          miRBase Alias:

          Mouse: mmu-miR-1a-1* (v17)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline