Search
Search

| Assay Name: | pma-miR-199a* |
|---|---|
| miRBase Accession Number: | MI0017101 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGUAGUCUGCACAUUGGUUA |
| Species: | Petromyzon marinus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000040, mir-199 |
Mature miRNA Sequence: | CAGUAGUCUGCACAUUGGUUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Petromyzon marinus | pma-miR-199a-3p | MIMAT0019506 |
| Petromyzon marinus | pma-miR-199b-3p | MIMAT0019508 |