Search
Search

| Assay Name: | rno-miR-222* |
|---|---|
| miRBase Accession Number: | MI0000962 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGCUCAGUAGCCAGUGUAGAU |
| Species: | Rat |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000051, mir-221 |
Mature miRNA Sequence: | GGCUCAGUAGCCAGUGUAGAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-222-5p | MIMAT0017164 |