Search
Search

| Assay Name: | mmu-miR-328* |
|---|---|
| miRBase Accession Number: | MI0000603 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGGGGGCAGGAGGGGCUCAGGG |
| Species: | Mouse, Rabbit |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000203, mir-328 |
Mature miRNA Sequence: | GGGGGGCAGGAGGGGCUCAGGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-328-5p | MIMAT0017030 |
| Rabbit | ocu-miR-328-5p | MIMAT0048376 |