Search
Search

| Assay Name: | hsa-miR-98-3p |
|---|---|
| miRBase Accession Number: | MI0000100 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUAUACAACUUACUACUUUCCC |
| Species: | Human, Goat, Rabbit |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | CUAUACAACUUACUACUUUCCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-98-3p | MIMAT0022842 |
| Goat | chi-miR-98-3p | MIMAT0036311 |
| Rabbit | ocu-miR-98-3p | MIMAT0048181 |