Search
Search

| Assay Name: | dre-miR-25-5p |
|---|---|
| miRBase Accession Number: | MI0001922 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGCGGAGACUUGGGCAGCUGCC |
| Species: | Zebrafish |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000013, mir-25 |
Mature miRNA Sequence: | AGGCGGAGACUUGGGCAGCUGCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Zebrafish | dre-miR-25-5p | MIMAT0031946 |