Search
Search

| Assay Name: | hsa-miR-211-3p |
|---|---|
| miRBase Accession Number: | MI0000287 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCAGGGACAGCAAAGGGGUGC |
| Species: | Human, Pteropus alecto, Rabbit |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000042, mir-204 |
Mature miRNA Sequence: | GCAGGGACAGCAAAGGGGUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-211-3p | MIMAT0022694 |
| Pteropus alecto | pal-miR-211-3p | MIMAT0040336 |
| Rabbit | ocu-miR-211-3p | MIMAT0048334 |