Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 477848_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-let-7d-3p
          Stem-loop Accession Number: MI0000065
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 94178834 - 94178920 [+] on Build GRCh38
          Mature miRNA Sequence: CUAUACGACCUGCUGCCUUUCU
          Species: Human, Mouse, Rat, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Pig, Rabbit
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 477848_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          CUAUACGACCUGCUGCCUUUCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7d-3p MIMAT0004484
          Mouse mmu-let-7d-3p MIMAT0000384
          Rat rno-let-7d-3p MIMAT0000563
          Cricetulus griseus cgr-let-7d-3p MIMAT0023720
          Dasypus novemcinctus dno-let-7d-3p MIMAT0047481
          Guinea pig cpo-let-7d-3p MIMAT0046825
          View More Rows
          Pig ssc-let-7d-3p MIMAT0025357
          Rabbit ocu-let-7d-3p MIMAT0048083

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7d
          Stem-loop Accession # MI0000065
          Stem-loop Sequence
          CCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCUUAGG
          Chromosome Location Chr. 9 - 94178834 - 94178920 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004484
          miRBase ID: hsa-let-7d-3p
          Mature miRNA Sequence: CUAUACGACCUGCUGCCUUUCU
          Chromosome Location: Chr. 9 - 94178834 - 94178920 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-let-7d
          Stem-loop Accession # MI0000405
          Stem-loop Sequence
          AAUGGGUUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGAGAUUUUGCCCACAAGGAGUUAACUAUACGACCUGCUGCCUUUCUUAGGGCCUUAUU
          Chromosome Location Chr. 13 - 48536012 - 48536114 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000384
          miRBase ID: mmu-let-7d-3p
          Mature miRNA Sequence: CUAUACGACCUGCUGCCUUUCU
          Chromosome Location: Chr. 13 - 48536012 - 48536114 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-let-7d
          Stem-loop Accession # MI0000601
          Stem-loop Sequence
          UGGGCUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGAGAUUUUGCCCACAAGGAGUUAACUAUACGACCUGCUGCCUUUCUUAGGGCCUU
          Chromosome Location Chr. 17 - 16419980 - 16420077 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000563
          miRBase ID: rno-let-7d-3p
          Mature miRNA Sequence: CUAUACGACCUGCUGCCUUUCU
          Chromosome Location: Chr. 17 - 16419980 - 16420077 [+] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-let-7d
          Stem-loop Accession # MI0020371
          Stem-loop Sequence
          GCUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCUUAGGGCCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023720
          miRBase ID: cgr-let-7d-3p
          Mature miRNA Sequence: CUAUACGACCUGCUGCCUUUCU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-let-7d
          Stem-loop Accession # MI0038890
          Stem-loop Sequence
          AGAGGUAGUAGGUUGCAUAGUUUUGGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047481
          miRBase ID: dno-let-7d-3p
          Mature miRNA Sequence: CUAUACGACCUGCUGCCUUUCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7d
          Stem-loop Accession # MI0038545
          Stem-loop Sequence
          AGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046825
          miRBase ID: cpo-let-7d-3p
          Mature miRNA Sequence: CUAUACGACCUGCUGCCUUUCU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-let-7d
          Stem-loop Accession # MI0022120
          Stem-loop Sequence
          GUUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGAGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCUUAGGGCCUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025357
          miRBase ID: ssc-let-7d-3p
          Mature miRNA Sequence: CUAUACGACCUGCUGCCUUUCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-let-7d
          Stem-loop Accession # MI0039210
          Stem-loop Sequence
          AGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048083
          miRBase ID: ocu-let-7d-3p
          Mature miRNA Sequence: CUAUACGACCUGCUGCCUUUCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465667_pri
          TaqMan™ Pri-miRNA Assay : Hs03302562_pri
          TaqMan™ Pri-miRNA Assay : Mm03306666_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10785
          Ambion® Pre-miR™ miRNA Precursor : PM10785
          TaqMan™ MicroRNA Assay : 001178
          mirVana® miRNA inhibitor : MH10785
          mirVana® miRNA mimic : MC10785
          TaqMan™ Advanced miRNA Assay : mmu481435_mir
          TaqMan™ Advanced miRNA Assay : rno481435_mir
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 1
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human B Card
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 1

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline