Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 477938_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-184
          Stem-loop Accession Number: MI0000481
          miRBase Version: v22.1
          Chromosome Location: Chr.15: 79209788 - 79209871 [+] on Build GRCh38
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Bovine, Callithrix jacchus, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Macaca nemestrina, Monodelphis domestica, Otolemur garnettii, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Taeniopygia guttata, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 477938_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000059, mir-184

          Mature miRNA Sequence:

          UGGACGGAGAACUGAUAAGGGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-184 MIMAT0000454
          Mouse mmu-miR-184-3p MIMAT0000213
          Rat rno-miR-184 MIMAT0000861
          Alligator mississippiensis ami-miR-184-3p MIMAT0038199
          Anolis carolinensis aca-miR-184-3p MIMAT0021814
          Bovine bta-miR-184 MIMAT0009246
          View More Rows
          Callithrix jacchus cja-miR-184 MIMAT0039492
          Chicken gga-miR-184-3p MIMAT0001158
          Chrysemys picta cpi-miR-184-3p MIMAT0037801
          Columba livia cli-miR-184-3p MIMAT0038561
          Cricetulus griseus cgr-miR-184 MIMAT0023809
          Dasypus novemcinctus dno-miR-184-3p MIMAT0047704
          Dog cfa-miR-184 MIMAT0009842
          Goat chi-miR-184 MIMAT0036009
          Guinea pig cpo-miR-184-3p MIMAT0047053
          Horse eca-miR-184 MIMAT0012894
          Macaca nemestrina mne-miR-184 MIMAT0002303
          Monodelphis domestica mdo-miR-184-3p MIMAT0004119
          Otolemur garnettii oga-miR-184 MIMAT0049718
          Pan troglodytes ptr-miR-184 MIMAT0002301
          Pig ssc-miR-184 MIMAT0002127
          Pongo pygmaeus ppy-miR-184 MIMAT0002302
          Pteropus alecto pal-miR-184-3p MIMAT0040165
          Rabbit ocu-miR-184-3p MIMAT0048290
          Rhesus monkey mml-miR-184 MIMAT0006218
          Taeniopygia guttata tgu-miR-184 MIMAT0014561
          Tupaia chinensis tch-miR-184 MIMAT0036493

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-184
          Stem-loop Accession # MI0000481
          Stem-loop Sequence
          CCAGUCACGUCCCCUUAUCACUUUUCCAGCCCAGCUUUGUGACUGUAAGUGUUGGACGGAGAACUGAUAAGGGUAGGUGAUUGA
          Chromosome Location Chr. 15 - 79209788 - 79209871 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000454
          miRBase ID: hsa-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: Chr. 15 - 79209788 - 79209871 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-184
          Stem-loop Accession # MI0000226
          Stem-loop Sequence
          CCUUUCCUUAUCACUUUUCCAGCCAGCUUUGUGACUCUAAGUGUUGGACGGAGAACUGAUAAGGGUAGG
          Chromosome Location Chr. 9 - 89802260 - 89802328 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000213
          miRBase ID: mmu-miR-184-3p
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: Chr. 9 - 89802260 - 89802328 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-184
          Stem-loop Accession # MI0000929
          Stem-loop Sequence
          CACUUUCCCUUAUCAGUUUUCCAGCCAGCUUUGUGACUGUAAAUGUUGGACGGAGAACUGAUAAGGGUAAGUGACUG
          Chromosome Location Chr. 8 - 97175657 - 97175733 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000861
          miRBase ID: rno-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: Chr. 8 - 97175657 - 97175733 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-184
          Stem-loop Accession # MI0029769
          Stem-loop Sequence
          ACGCCAUUCCCAUCUCCUUAUCACUUUUCCAGCCCAGCUUUCUCAUGCUCACUGUUGGACGGAGAACUGAUAAGGGUAUGUGACUGACACGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038199
          miRBase ID: ami-miR-184-3p
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-184
          Stem-loop Accession # MI0018781
          Stem-loop Sequence
          CUGGAGCCGCUCACGUCCCCUUAUCACUUUUCCAGCCAGCUUUCUUGUCAUAACCGUUGGACGGAGAACUGAUAAGGGUUUGUGGGUGAGACCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021814
          miRBase ID: aca-miR-184-3p
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-184
          Stem-loop Accession # MI0009757
          Stem-loop Sequence
          CCAGUCACAUCCCCUUAUCACUUUUCCAGCCAGCUUUGUGACUCUAACUGUUGGACGGAGAACUGAUAAGGGUAGGUGAUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009246
          miRBase ID: bta-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-184
          Stem-loop Accession # MI0032017
          Stem-loop Sequence
          CAGUCACGUCCCCUUAUCACUUUUCCAGCCCAGCUUUGUGACUGUAAGUGUUGGACGGAGAACUGAUAAGGGUAGGUGACUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039492
          miRBase ID: cja-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-184
          Stem-loop Accession # MI0001227
          Stem-loop Sequence
          CCGCUCUCACCCCCUUAUCACUUUUCCAGCCCAGCUUCUUCGCUCUGACUGUUGGACGGAGAACUGAUAAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001158
          miRBase ID: gga-miR-184-3p
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-184
          Stem-loop Accession # MI0029508
          Stem-loop Sequence
          GCAGCCAUUCUCGUGUCCUUAUCACUUUUCCAGCCCAGCUUUUUCAUUGUAACUGUUGGACGGAGAACUGAUAAGGGUAUGUGACUGACAUGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037801
          miRBase ID: cpi-miR-184-3p
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-184
          Stem-loop Accession # MI0029995
          Stem-loop Sequence
          GGCCAUUCUCAUCUCCUUAUCACUUUUCCAGCCCAGCUUCUUCACUGUAACUGUUGGACGGAGAACUGAUAAGGGUAUGCGAGUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038561
          miRBase ID: cli-miR-184-3p
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-184
          Stem-loop Accession # MI0020427
          Stem-loop Sequence
          UCGCCUGGCAAUCACGUUUCCUUAUCACUUUUCCAGCCAGCUUUGUGACUCUAAGUGUUGGACGGAGAACUGAUAAGGGUAGGUGAUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023809
          miRBase ID: cgr-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-184
          Stem-loop Accession # MI0039015
          Stem-loop Sequence
          CCCUUAUCACUUUUCCAGCCCAGCUUUCUAACUCGAAGUGUUGGACGGAGAACUGAUAAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047704
          miRBase ID: dno-miR-184-3p
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-184
          Stem-loop Accession # MI0010337
          Stem-loop Sequence
          CCCCUUAUCACUUUUCCAGCCAGCUUUGUGACUCUAAGUGUUGGACGGAGAACUGAUAAGGGUAGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009842
          miRBase ID: cfa-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-184
          Stem-loop Accession # MI0030656
          Stem-loop Sequence
          CCAGUCACAUCCCCUUAUCACUUUUCCAGCCAGCUUUGUGACUCUAACUGUUGGACGGAGAACUGAUAAGGGUAGGUGAUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036009
          miRBase ID: chi-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-184
          Stem-loop Accession # MI0038672
          Stem-loop Sequence
          CCUUAUCACUUUUCCAGCCCAGCUUUGUGAUUGUCAGUGUUGGACGGAGAACUGAUAAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047053
          miRBase ID: cpo-miR-184-3p
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-184
          Stem-loop Accession # MI0012650
          Stem-loop Sequence
          ACAUCUCCUUAUCACUUUUCCAGCCAGCUUUGUGACUCUAAGUGUUGGACGGAGAACUGAUAAGGGUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012894
          miRBase ID: eca-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-184
          Stem-loop Accession # MI0002604
          Stem-loop Sequence
          UCAGUCACGUCCCCUUAUCACUUUUCCAGCCCAGCUUUAUGACUGUAAGUGUUGGACGGAGAACUGAUAAGGGUAGGUGAUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002303
          miRBase ID: mne-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-184
          Stem-loop Accession # MI0005308
          Stem-loop Sequence
          CCAGUCACAUCCCCUUAUCACUUUUCCAGCCCAGCUUUCUAAUGCUAAUUGUUGGACGGAGAACUGAUAAGGGUAGGUGAUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004119
          miRBase ID: mdo-miR-184-3p
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-184
          Stem-loop Accession # MI0040510
          Stem-loop Sequence
          CCUUAUCACUUUUCCAGCCCAGCUGUGUGACUCUAGUGUUGGACGGAGAACUGAUAAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049718
          miRBase ID: oga-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-184
          Stem-loop Accession # MI0002602
          Stem-loop Sequence
          CCAGUCACGUCCCCUUAUCACUUUUCCAGCCCAGCUUUGUGACUGUAAGUGUUGGACGGAGAACUGAUAAGGGUAGGUGAUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002301
          miRBase ID: ptr-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-184
          Stem-loop Accession # MI0002421
          Stem-loop Sequence
          CCAGUCACAUCCCCUUAUCACUUUUCCAGCCAGCUUUGUGACUCUAAAUGUUGGACGGAGAACUGAUAAGGGUAGGUGAUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002127
          miRBase ID: ssc-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-184
          Stem-loop Accession # MI0002603
          Stem-loop Sequence
          CCAGUCACGUCCCCUUAUCACUUUUCCAGCCCAGCUUUGUGACUGUAAGUGUUGGACGGAGAACUGAUAAGGGUAGGUGAUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002302
          miRBase ID: ppy-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-184
          Stem-loop Accession # MI0032528
          Stem-loop Sequence
          CCUUAUCACUUUUCCAGCCAGCUUCGUGAUGCUCAGUGUUGGACGGAGAACUGAUAAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040165
          miRBase ID: pal-miR-184-3p
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-184
          Stem-loop Accession # MI0039323
          Stem-loop Sequence
          CCCUUAUCACUUUUCCAGCCCAGCUUUGUGACUCUGACUGUUGGACGGAGAACUGAUAAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048290
          miRBase ID: ocu-miR-184-3p
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-184
          Stem-loop Accession # MI0007648
          Stem-loop Sequence
          UCAGUCACGUCCCCUUAUCACUUUUCCAGCCCAGCUUUAUGACUGUAAGUGUUGGACGGAGAACUGAUAAGGGUAGGUGAUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006218
          miRBase ID: mml-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-184
          Stem-loop Accession # MI0013779
          Stem-loop Sequence
          CCCGCGGCUCCCCGCCGGUCUCCUGAGGCCGCUCUCGUGCCCUUAUCACUUUUCCAGCCCAGCUUCUGCACUCUGACUGUUGGACGGAGAACUGAUAAGGGUGCGCGAGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014561
          miRBase ID: tgu-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-184
          Stem-loop Accession # MI0031150
          Stem-loop Sequence
          CCUUAUCACUUUUCCAGCCCAGCUUUGUGACUCUGAGUGUUGGACGGAGAACUGAUAAGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036493
          miRBase ID: tch-miR-184
          Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303283_pri
          TaqMan™ Pri-miRNA Assay : Mm03306435_pri
          TaqMan™ Pri-miRNA Assay : Rn03465331_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10207
          Ambion® Pre-miR™ miRNA Precursor : PM10207
          TaqMan™ MicroRNA Assay : 000485
          mirVana® miRNA inhibitor : MH10207
          mirVana® miRNA mimic : MC10207
          TaqMan™ Advanced miRNA Assay : mmu480962_mir
          TaqMan™ Advanced miRNA Assay : rno480962_mir
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® Advanced miRNA Human A Card
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline