Search
Search

| Assay Name: | hsa-miR-412-3p |
|---|---|
| Stem-loop Accession Number: | MI0002464 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACUUCACCUGGUCCACUAGCCGU |
| Species: | Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000192, mir-412 |
Mature miRNA Sequence: | ACUUCACCUGGUCCACUAGCCGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-412-3p | MIMAT0002170 |
| Pan troglodytes | ptr-miR-412 | MIMAT0008139 |
| Pongo pygmaeus | ppy-miR-412 | MIMAT0015879 |
| Rhesus monkey | mml-miR-412-3p | MIMAT0006318 |