Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478794_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-29b-1-5p
          Stem-loop Accession Number: MI0000105
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 130877459 - 130877539 [-] on Build GRCh38
          Mature miRNA Sequence: GCUGGUUUCAUAUGGUGGUUUAGA
          Species: Human, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Monodelphis domestica, Rabbit
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478794_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000009, mir-29

          Mature miRNA Sequence:

          GCUGGUUUCAUAUGGUGGUUUAGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-29b-1-5p MIMAT0004514
          Columba livia cli-miR-29b-5p MIMAT0038449
          Cricetulus griseus cgr-miR-29b-5p MIMAT0023885
          Dasypus novemcinctus dno-miR-29b-5p MIMAT0047551
          Guinea pig cpo-miR-29b-5p MIMAT0046898
          Monodelphis domestica mdo-miR-29b-1-5p MIMAT0026703
          View More Rows
          Rabbit ocu-miR-29b-5p MIMAT0048145

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-29b-1
          Stem-loop Accession # MI0000105
          Stem-loop Sequence
          CUUCAGGAAGCUGGUUUCAUAUGGUGGUUUAGAUUUAAAUAGUGAUUGUCUAGCACCAUUUGAAAUCAGUGUUCUUGGGGG
          Chromosome Location Chr. 7 - 130877459 - 130877539 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004514
          miRBase ID: hsa-miR-29b-1-5p
          Mature miRNA Sequence: GCUGGUUUCAUAUGGUGGUUUAGA
          Chromosome Location: Chr. 7 - 130877459 - 130877539 [-] on Build GRCh38

          Columba livia

          Stem-loop ID cli-mir-29b-1
          Stem-loop Accession # MI0029928
          Stem-loop Sequence
          CCUCAGGAAGCUGGUUUCAUAUGGUGGUUUAGAUUUAACUAUUCAUUGUCUAGCACCAUUUGAAAUCAGUGUUCUUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038449
          miRBase ID: cli-miR-29b-5p
          Mature miRNA Sequence: GCUGGUUUCAUAUGGUGGUUUAGA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-29b
          Stem-loop Accession # MI0020479
          Stem-loop Sequence
          CCUUCAGGAAGCUGGUUUCAUAUGGUGGUUUAGAUUUAAAUAGUGAUUGUCUAGCACCAUUUGAAAUCAGUGUUCUUGGUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023885
          miRBase ID: cgr-miR-29b-5p
          Mature miRNA Sequence: GCUGGUUUCAUAUGGUGGUUUAGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-29b-1
          Stem-loop Accession # MI0038931
          Stem-loop Sequence
          GCUGGUUUCAUAUGGUGGUUUAGAUUUAAAUAGUGAUUGUCUAGCACCAUUUGAAAUCAGUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047551
          miRBase ID: dno-miR-29b-5p
          Mature miRNA Sequence: GCUGGUUUCAUAUGGUGGUUUAGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-29b-1
          Stem-loop Accession # MI0038587
          Stem-loop Sequence
          GCUGGUUUCAUAUGGUGGUUUAGAUUUAAAUAGUGAUUGUCUAGCACCAUUUGAAAUCAGUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046898
          miRBase ID: cpo-miR-29b-5p
          Mature miRNA Sequence: GCUGGUUUCAUAUGGUGGUUUAGA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-29b-1
          Stem-loop Accession # MI0005352
          Stem-loop Sequence
          GCUUCUUCAGGAAGCUGGUUUCAUAUGGUGGUUUAGAUUUAACUACUGAGUGUCUAGCACCAUUUGAAAUCAGUGUUCUUGGAGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026703
          miRBase ID: mdo-miR-29b-1-5p
          Mature miRNA Sequence: GCUGGUUUCAUAUGGUGGUUUAGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-29b-1
          Stem-loop Accession # MI0039244
          Stem-loop Sequence
          GCUGGUUUCAUAUGGUGGUUUAGAUUUAAAUAGUGAUUGUCUAGCACCAUUUGAAAUCAGUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048145
          miRBase ID: ocu-miR-29b-5p
          Mature miRNA Sequence: GCUGGUUUCAUAUGGUGGUUUAGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302748_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12434
          Ambion® Pre-miR™ miRNA Precursor : PM12434
          TaqMan™ MicroRNA Assay : 002165
          mirVana® miRNA inhibitor : MH12434
          mirVana® miRNA mimic : MC12434
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human B Card
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-cvz79:80/100.66.78.224:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0