Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 479611_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-3120-5p
          Stem-loop Accession Number: MI0014136
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 172138808 - 172138888 [+] on Build GRCh38
          Mature miRNA Sequence: CCUGUCUGUGCCUGCUGUACA
          Species: Human, Ophiophagus hannah
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 479611_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000062, mir-214

          Mature miRNA Sequence:

          CCUGUCUGUGCCUGCUGUACA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-3120-5p MIMAT0019198
          Ophiophagus hannah oha-miR-3120 MIMAT0036830

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-3120
          Stem-loop Accession # MI0014136
          Stem-loop Sequence
          GUCAUGUGACUGCCUGUCUGUGCCUGCUGUACAGGUGAGCGGAUGUUCUGCACAGCAAGUGUAGACAGGCAGACACAUGAC
          Chromosome Location Chr. 1 - 172138808 - 172138888 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0019198
          miRBase ID: hsa-miR-3120-5p
          Mature miRNA Sequence: CCUGUCUGUGCCUGCUGUACA
          Chromosome Location: Chr. 1 - 172138808 - 172138888 [+] on Build GRCh38

          Ophiophagus hannah

          Stem-loop ID oha-mir-3120
          Stem-loop Accession # MI0031415
          Stem-loop Sequence
          GGAUUGUCAUGUGACUGCCUGUCUGUGCCUGCUGUACAGGUGAGAGGAUGUUCUGCACAGCAAGUGUAGACAGGCAGACACAUGACAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036830
          miRBase ID: oha-miR-3120
          Mature miRNA Sequence: CCUGUCUGUGCCUGCUGUACA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302971_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM21017
          Ambion® Pre-miR™ miRNA Precursor : PM21017
          TaqMan™ MicroRNA Assay : 462016_mat
          mirVana® miRNA inhibitor : MH21017
          mirVana® miRNA mimic : MC21017

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline