Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12795
          Assay Name: hsa-miR-26a-2-3p
          miRBase Accession Number: MI0000750
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 57824609 - 57824692 [-] on Build GRCh38
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Species: Human, Chrysemys picta, Dasypus novemcinctus, Guinea pig, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12795
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000043, mir-26

          Mature miRNA Sequence:

          CCUAUUCUUGAUUACUUGUUUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-26a-2-3p MIMAT0004681
          Chrysemys picta cpi-miR-26-2-3p MIMAT0037662
          Dasypus novemcinctus dno-miR-26a-2-3p MIMAT0047540
          Guinea pig cpo-miR-26a-2-3p MIMAT0046887
          Pteropus alecto pal-miR-26a-2-3p MIMAT0039804
          Python bivittatus pbv-miR-26-3-3p MIMAT0038860
          View More Rows
          Rabbit ocu-miR-26a-3p MIMAT0048136
          Rhesus monkey mml-miR-26a-2-3p MIMAT0031062
          Salmo salar ssa-miR-26a-4-3p MIMAT0032566

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-26a-2
          Stem-loop Accession # MI0000750
          Stem-loop Sequence
          GGCUGUGGCUGGAUUCAAGUAAUCCAGGAUAGGCUGUUUCCAUCUGUGAGGCCUAUUCUUGAUUACUUGUUUCUGGAGGCAGCU
          Chromosome Location Chr. 12 - 57824609 - 57824692 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004681
          miRBase ID: hsa-miR-26a-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: Chr. 12 - 57824609 - 57824692 [-] on Build GRCh38

          Chrysemys picta

          Stem-loop ID cpi-mir-26-2
          Stem-loop Accession # MI0029416
          Stem-loop Sequence
          UGCGGAGGCCGUAGCCCGGUUCAAGUAAUCCAGGAUAGGCUGUUACCAGGCAGCACGGCCUAUUCUUGAUUACUUGUUUCAGGAGGCGGCUGCUGGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037662
          miRBase ID: cpi-miR-26-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-26a-2
          Stem-loop Accession # MI0038925
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGUUUCCAUCCGCGAGGCCUAUUCUUGAUUACUUGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047540
          miRBase ID: dno-miR-26a-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-26a-2
          Stem-loop Accession # MI0038581
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGUUUCCAUCUGUGAGGCCUAUUCUUGAUUACUUGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046887
          miRBase ID: cpo-miR-26a-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-26a-2
          Stem-loop Accession # MI0032337
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGUUUCCAUCUGUGAGGCCUAUUCUUGAUUACUUGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039804
          miRBase ID: pal-miR-26a-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-26-3
          Stem-loop Accession # MI0030176
          Stem-loop Sequence
          GCGAAGGUCUCAGCCUGGUUCAAGUAAUCCAGGAUAGGCUGUUACCAGGCAACACGGCCUAUUCUUGAUUACUUGUUUCAGGAGGCGGCUUCUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038860
          miRBase ID: pbv-miR-26-3-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-26a
          Stem-loop Accession # MI0039239
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGUUGCCAUCCGUGAGGCCUAUUCUUGAUUACUUGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048136
          miRBase ID: ocu-miR-26a-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-26a-2
          Stem-loop Accession # MI0007593
          Stem-loop Sequence
          GGCUGUGGCUGGAUUCAAGUAAUCCAGGAUAGGCUGUUUCCAUCUGUGAGGCCUAUUCUUGAUUACUUGUUUCUGGAGGCAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031062
          miRBase ID: mml-miR-26a-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-26a-4
          Stem-loop Accession # MI0026652
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGGUUAACACCUGUACGGCCUAUUCUUGAUUACUUGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032566
          miRBase ID: ssa-miR-26a-4-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303430_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12795
          TaqMan™ MicroRNA Assay : 002115
          mirVana® miRNA inhibitor : MH12795
          mirVana® miRNA mimic : MC12795
          TaqMan™ Advanced miRNA Assay : 478788_mir


          miRBase Alias:

          Human: hsa-miR-26a-2* (v17)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline