Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478788_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-26a-2-3p
          Stem-loop Accession Number: MI0000750
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 57824609 - 57824692 [-] on Build GRCh38
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Species: Human, Chrysemys picta, Dasypus novemcinctus, Guinea pig, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478788_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000043, mir-26

          Mature miRNA Sequence:

          CCUAUUCUUGAUUACUUGUUUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-26a-2-3p MIMAT0004681
          Chrysemys picta cpi-miR-26-2-3p MIMAT0037662
          Dasypus novemcinctus dno-miR-26a-2-3p MIMAT0047540
          Guinea pig cpo-miR-26a-2-3p MIMAT0046887
          Pteropus alecto pal-miR-26a-2-3p MIMAT0039804
          Python bivittatus pbv-miR-26-3-3p MIMAT0038860
          View More Rows
          Rabbit ocu-miR-26a-3p MIMAT0048136
          Rhesus monkey mml-miR-26a-2-3p MIMAT0031062
          Salmo salar ssa-miR-26a-4-3p MIMAT0032566

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-26a-2
          Stem-loop Accession # MI0000750
          Stem-loop Sequence
          GGCUGUGGCUGGAUUCAAGUAAUCCAGGAUAGGCUGUUUCCAUCUGUGAGGCCUAUUCUUGAUUACUUGUUUCUGGAGGCAGCU
          Chromosome Location Chr. 12 - 57824609 - 57824692 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004681
          miRBase ID: hsa-miR-26a-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: Chr. 12 - 57824609 - 57824692 [-] on Build GRCh38

          Chrysemys picta

          Stem-loop ID cpi-mir-26-2
          Stem-loop Accession # MI0029416
          Stem-loop Sequence
          UGCGGAGGCCGUAGCCCGGUUCAAGUAAUCCAGGAUAGGCUGUUACCAGGCAGCACGGCCUAUUCUUGAUUACUUGUUUCAGGAGGCGGCUGCUGGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037662
          miRBase ID: cpi-miR-26-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-26a-2
          Stem-loop Accession # MI0038925
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGUUUCCAUCCGCGAGGCCUAUUCUUGAUUACUUGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047540
          miRBase ID: dno-miR-26a-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-26a-2
          Stem-loop Accession # MI0038581
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGUUUCCAUCUGUGAGGCCUAUUCUUGAUUACUUGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046887
          miRBase ID: cpo-miR-26a-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-26a-2
          Stem-loop Accession # MI0032337
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGUUUCCAUCUGUGAGGCCUAUUCUUGAUUACUUGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039804
          miRBase ID: pal-miR-26a-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-26-3
          Stem-loop Accession # MI0030176
          Stem-loop Sequence
          GCGAAGGUCUCAGCCUGGUUCAAGUAAUCCAGGAUAGGCUGUUACCAGGCAACACGGCCUAUUCUUGAUUACUUGUUUCAGGAGGCGGCUUCUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038860
          miRBase ID: pbv-miR-26-3-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-26a
          Stem-loop Accession # MI0039239
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGUUGCCAUCCGUGAGGCCUAUUCUUGAUUACUUGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048136
          miRBase ID: ocu-miR-26a-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-26a-2
          Stem-loop Accession # MI0007593
          Stem-loop Sequence
          GGCUGUGGCUGGAUUCAAGUAAUCCAGGAUAGGCUGUUUCCAUCUGUGAGGCCUAUUCUUGAUUACUUGUUUCUGGAGGCAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031062
          miRBase ID: mml-miR-26a-2-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-26a-4
          Stem-loop Accession # MI0026652
          Stem-loop Sequence
          UUCAAGUAAUCCAGGAUAGGCUGGUUAACACCUGUACGGCCUAUUCUUGAUUACUUGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032566
          miRBase ID: ssa-miR-26a-4-3p
          Mature miRNA Sequence: CCUAUUCUUGAUUACUUGUUUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303430_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12795
          Ambion® Pre-miR™ miRNA Precursor : PM12795
          TaqMan™ MicroRNA Assay : 002115
          mirVana® miRNA inhibitor : MH12795
          mirVana® miRNA mimic : MC12795
          FGMIR : TaqManTM Advanced miRNA Human B Card
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline