Search
Search

| Assay Name: | hsa-let-7e-3p |
|---|---|
| miRBase Accession Number: | MI0000066 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUAUACGGCCUCCUAGCUUUCC |
| Species: | Human, Mouse, Rat, Cricetulus griseus, Goat, Guinea pig, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | CUAUACGGCCUCCUAGCUUUCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-let-7e-3p | MIMAT0004485 |
| Mouse | mmu-let-7e-3p | MIMAT0017016 |
| Rat | rno-let-7e-3p | MIMAT0004706 |
| Cricetulus griseus | cgr-let-7e | MIMAT0041132 |
| Goat | chi-let-7e-3p | MIMAT0035889 |
| Guinea pig | cpo-let-7e-3p | MIMAT0046827 |
| View More Rows | ||
| Rhesus monkey | mml-let-7e-3p | MIMAT0026792 |