Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03465947_pri
          Assay Name:
          rno-let-7e
          Stem-loop ID:
          rno-let-7e rno-mir-125a
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.1: 59704381 - 59704473 [+] on Build Rnor_6.0
          Stem-loop Location:
          Chr.1: 59704381 - 59704473 [+] on Build Rnor_6.0 Chr.1: 59704827 - 59704911 [+] on Build Rnor_6.0
          Mature miRNA ID:
          rno-let-7e-3p rno-let-7e-5p rno-miR-125a-3p rno-miR-125a-5p
          Species:
          Rat
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Rn03465947_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          116

          Chromosome Location:

          Chr. 1 - 59704631 - 59704631 [+] on 59704631 Build Rnor_6.0

          Species:

          Rat

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-let-7e-3p MIMAT0004706
          Rat rno-let-7e-5p MIMAT0000777
          Rat rno-miR-125a-3p MIMAT0004729
          Rat rno-miR-125a-5p MIMAT0000829

          Stem-loop Details



          Rat

          Stem-loop ID rno-let-7e
          Stem-loop Accession # MI0000832
          Stem-loop Sequence
          CGCGCCCCCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAAGACACCCGAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGGCUGCGCC
          Chromosome Location Chr. 1 - 59704381 - 59704473 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004706
          miRBase ID: rno-let-7e-3p
          Mature miRNA Sequence: CUAUACGGCCUCCUAGCUUUCC
          Chromosome Location: Chr. 1 - 59704381 - 59704473 [+] on Build Rnor_6.0

          miRBase Accession #: MIMAT0000777
          miRBase ID: rno-let-7e-5p
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: Chr. 1 - 59704381 - 59704473 [+] on Build Rnor_6.0

          Gene Family ID MIPF0000002, let-7
          Distance to Amplicon 104 bases
          Stem-loop ID rno-mir-125a
          Stem-loop Accession # MI0000895
          Stem-loop Sequence
          UGCCGGCCUCUGGGUCCCUGAGACCCUUUAACCUGUGAGGACGUCCAGGGUCACAGGUGAGGUUCUUGGGAGCCUGGCGCCUGGC
          Chromosome Location Chr. 1 - 59704827 - 59704911 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004729
          miRBase ID: rno-miR-125a-3p
          Mature miRNA Sequence: ACAGGUGAGGUUCUUGGGAGCC
          Chromosome Location: Chr. 1 - 59704827 - 59704911 [+] on Build Rnor_6.0

          miRBase Accession #: MIMAT0000829
          miRBase ID: rno-miR-125a-5p
          Mature miRNA Sequence: UCCCUGAGACCCUUUAACCUGUGA
          Chromosome Location: Chr. 1 - 59704827 - 59704911 [+] on Build Rnor_6.0

          Gene Family ID MIPF0000033, mir-10
          Distance to Amplicon 135 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM12378
          Ambion® Pre-miR™ miRNA Precursor : PM12378
          TaqMan™ MicroRNA Assay : 002199
          mirVana® miRNA inhibitor : MH12378
          mirVana® miRNA mimic : MC12378
          TaqMan™ Advanced miRNA Assay : 477883_mir
          TaqMan™ Advanced miRNA Assay : mmu481497_mir
          TaqMan™ Advanced miRNA Assay : rno481302_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12855
          Ambion® Pre-miR™ miRNA Precursor : PM12855
          TaqMan™ MicroRNA Assay : 002407
          mirVana® miRNA inhibitor : MH12855
          mirVana® miRNA mimic : MC12855
          TaqMan™ Advanced miRNA Assay : rno479281_mir
          TaqMan™ Advanced miRNA Assay : 479281_mir
          TaqMan™ Advanced miRNA Assay : mmu483017_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12561
          Ambion® Pre-miR™ miRNA Precursor : PM12561
          TaqMan™ MicroRNA Assay : 002198
          mirVana® miRNA inhibitor : MH12561
          mirVana® miRNA mimic : MC12561
          TaqMan™ Advanced miRNA Assay : rno480906_mir
          TaqMan™ Advanced miRNA Assay : 477884_mir
          TaqMan™ Advanced miRNA Assay : mmu480906_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12304
          Ambion® Pre-miR™ miRNA Precursor : PM12304
          TaqMan™ MicroRNA Assay : 002406
          mirVana® miRNA inhibitor : MH12304
          mirVana® miRNA mimic : MC12304
          TaqMan™ Advanced miRNA Assay : 478579_mir


          miRBase Alias:

          Rat: rno-mir-125a (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline