Search
Search

| Assay Name: | hsa-let-7e-5p |
|---|---|
| Stem-loop Accession Number: | MI0000066 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGGUAGGAGGUUGUAUAGUU |
| Species: | Human, Mouse, Rat, Callithrix jacchus, Dog, Goat, Guinea pig, Horse, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Saimiri boliviensis, Tupaia chinensis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | UGAGGUAGGAGGUUGUAUAGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-let-7e-5p | MIMAT0000066 |
| Mouse | mmu-let-7e-5p | MIMAT0000524 |
| Rat | rno-let-7e-5p | MIMAT0000777 |
| Callithrix jacchus | cja-let-7e | MIMAT0049312 |
| Dog | cfa-let-7e | MIMAT0006608 |
| Goat | chi-let-7e-5p | MIMAT0035888 |
| View More Rows | ||
| Guinea pig | cpo-let-7e-5p | MIMAT0046826 |
| Horse | eca-let-7e | MIMAT0013007 |
| Nomascus leucogenys | nle-let-7e | MIMAT0049411 |
| Otolemur garnettii | oga-let-7e | MIMAT0049652 |
| Pan paniscus | ppa-let-7e | MIMAT0049090 |
| Pan troglodytes | ptr-let-7e | MIMAT0007940 |
| Papio hamadryas | pha-let-7e | MIMAT0049516 |
| Pig | ssc-let-7e | MIMAT0013866 |
| Pongo pygmaeus | ppy-let-7e | MIMAT0015725 |
| Pteropus alecto | pal-let-7e-5p | MIMAT0039784 |
| Rhesus monkey | mml-let-7e-5p | MIMAT0006155 |
| Saimiri boliviensis | sbo-let-7e | MIMAT0049471 |
| Tupaia chinensis | tch-let-7e-5p | MIMAT0036620 |