Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12304
          Assay Name: hsa-let-7e-5p
          miRBase Accession Number: MI0000066
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 51692786 - 51692864 [+] on Build GRCh38
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Species: Human, Mouse, Rat, Callithrix jacchus, Dog, Goat, Guinea pig, Horse, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey, Saimiri boliviensis, Tupaia chinensis
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12304
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          UGAGGUAGGAGGUUGUAUAGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7e-5p MIMAT0000066
          Mouse mmu-let-7e-5p MIMAT0000524
          Rat rno-let-7e-5p MIMAT0000777
          Callithrix jacchus cja-let-7e MIMAT0049312
          Dog cfa-let-7e MIMAT0006608
          Goat chi-let-7e-5p MIMAT0035888
          View More Rows
          Guinea pig cpo-let-7e-5p MIMAT0046826
          Horse eca-let-7e MIMAT0013007
          Nomascus leucogenys nle-let-7e MIMAT0049411
          Otolemur garnettii oga-let-7e MIMAT0049652
          Pan paniscus ppa-let-7e MIMAT0049090
          Pan troglodytes ptr-let-7e MIMAT0007940
          Papio hamadryas pha-let-7e MIMAT0049516
          Pig ssc-let-7e MIMAT0013866
          Pongo pygmaeus ppy-let-7e MIMAT0015725
          Pteropus alecto pal-let-7e-5p MIMAT0039784
          Rhesus monkey mml-let-7e-5p MIMAT0006155
          Saimiri boliviensis sbo-let-7e MIMAT0049471
          Tupaia chinensis tch-let-7e-5p MIMAT0036620

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7e
          Stem-loop Accession # MI0000066
          Stem-loop Sequence
          CCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGG
          Chromosome Location Chr. 19 - 51692786 - 51692864 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000066
          miRBase ID: hsa-let-7e-5p
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: Chr. 19 - 51692786 - 51692864 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-let-7e
          Stem-loop Accession # MI0000561
          Stem-loop Sequence
          CGCGCCCCCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAAGACACCCGAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGGCUGCGCC
          Chromosome Location Chr. 17 - 17830352 - 17830444 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000524
          miRBase ID: mmu-let-7e-5p
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: Chr. 17 - 17830352 - 17830444 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-let-7e
          Stem-loop Accession # MI0000832
          Stem-loop Sequence
          CGCGCCCCCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAAGACACCCGAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGGCUGCGCC
          Chromosome Location Chr. 1 - 59704381 - 59704473 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000777
          miRBase ID: rno-let-7e-5p
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: Chr. 1 - 59704381 - 59704473 [+] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-let-7e
          Stem-loop Accession # MI0040048
          Stem-loop Sequence
          UGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGACCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049312
          miRBase ID: cja-let-7e
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-let-7e
          Stem-loop Accession # MI0008004
          Stem-loop Sequence
          UGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCACGGAGAUCACUAUACGGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006608
          miRBase ID: cfa-let-7e
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-let-7e
          Stem-loop Accession # MI0030583
          Stem-loop Sequence
          ACCUGCCGCGCCCCCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGGCUGCGCCCUGCACGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035888
          miRBase ID: chi-let-7e-5p
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7e
          Stem-loop Accession # MI0038546
          Stem-loop Sequence
          UGAGGUAGGAGGUUGUAUAGUUGAGAAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046826
          miRBase ID: cpo-let-7e-5p
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-let-7e
          Stem-loop Accession # MI0012759
          Stem-loop Sequence
          CCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCACGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013007
          miRBase ID: eca-let-7e
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-let-7e
          Stem-loop Accession # MI0040151
          Stem-loop Sequence
          UGAGGUAGGAGGUUGUAUAGUUGAGGAAGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049411
          miRBase ID: nle-let-7e
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-let-7e
          Stem-loop Accession # MI0040435
          Stem-loop Sequence
          UGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049652
          miRBase ID: oga-let-7e
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-let-7e
          Stem-loop Accession # MI0039799
          Stem-loop Sequence
          UGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049090
          miRBase ID: ppa-let-7e
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-let-7e
          Stem-loop Accession # MI0008403
          Stem-loop Sequence
          CCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007940
          miRBase ID: ptr-let-7e
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-let-7e
          Stem-loop Accession # MI0040281
          Stem-loop Sequence
          UGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049516
          miRBase ID: pha-let-7e
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-let-7e
          Stem-loop Accession # MI0013086
          Stem-loop Sequence
          GCCCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCACGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013866
          miRBase ID: ssc-let-7e
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-let-7e
          Stem-loop Accession # MI0014785
          Stem-loop Sequence
          CCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015725
          miRBase ID: ppy-let-7e
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-let-7e
          Stem-loop Accession # MI0032326
          Stem-loop Sequence
          UGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCACGGAGAUCACUAUACGGCCUCCUAGCUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039784
          miRBase ID: pal-let-7e-5p
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-let-7e
          Stem-loop Accession # MI0007576
          Stem-loop Sequence
          CCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006155
          miRBase ID: mml-let-7e-5p
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Saimiri boliviensis

          Stem-loop ID sbo-let-7e
          Stem-loop Accession # MI0040228
          Stem-loop Sequence
          UGAGGUAGGAGGUUGUAUAGUUGAGCAGGACACCCAAGGAGAUCACUAUACGACCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049471
          miRBase ID: sbo-let-7e
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-let-7e
          Stem-loop Accession # MI0031293
          Stem-loop Sequence
          UGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036620
          miRBase ID: tch-let-7e-5p
          Mature miRNA Sequence: UGAGGUAGGAGGUUGUAUAGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465947_pri
          TaqMan™ Pri-miRNA Assay : Mm03306232_pri
          TaqMan™ Pri-miRNA Assay : Hs03295173_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12304
          TaqMan™ MicroRNA Assay : 002406
          mirVana® miRNA inhibitor : MH12304
          mirVana® miRNA mimic : MC12304
          TaqMan™ Advanced miRNA Assay : 478579_mir


          miRBase Alias:

          Human: hsa-let-7e (v17)
          Mouse: mmu-let-7e (v17)
          Rat: rno-let-7e (v18)
          Macaca mulatta: mml-let-7e (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline