Search
Search

| Assay Name: | hsa-miR-125a-3p |
|---|---|
| miRBase Accession Number: | MI0000469 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACAGGUGAGGUUCUUGGGAGCC |
| Species: | Human, Mouse, Rat, Guinea pig, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | ACAGGUGAGGUUCUUGGGAGCC |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-125a-3p | MIMAT0004602 | |
| Mouse | mmu-miR-125a-3p | MIMAT0004528 | |
| Rat | rno-miR-125a-3p | MIMAT0004729 | |
| Guinea pig | cpo-miR-125a-3p | MIMAT0046966 | |
| Horse | eca-miR-125a-3p | MIMAT0013009 | |
| Pan troglodytes | ptr-miR-125a | MIMAT0007984 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-125a-3p | MIMAT0015749 | |
| Rhesus monkey | mml-miR-125a-3p | MIMAT0006182 | |