Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481302_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-125a-3p
          Stem-loop Accession Number: MI0000469
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 51693254 - 51693339 [+] on Build GRCh38
          Mature miRNA Sequence: ACAGGUGAGGUUCUUGGGAGCC
          Species: Human, Mouse, Rat, Guinea pig, Horse, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481302_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          ACAGGUGAGGUUCUUGGGAGCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-125a-3p MIMAT0004602
          Mouse mmu-miR-125a-3p MIMAT0004528
          Rat rno-miR-125a-3p MIMAT0004729
          Guinea pig cpo-miR-125a-3p MIMAT0046966
          Horse eca-miR-125a-3p MIMAT0013009
          Pan troglodytes ptr-miR-125a MIMAT0007984
          View More Rows
          Pongo pygmaeus ppy-miR-125a-3p MIMAT0015749
          Rhesus monkey mml-miR-125a-3p MIMAT0006182

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-125a
          Stem-loop Accession # MI0000469
          Stem-loop Sequence
          UGCCAGUCUCUAGGUCCCUGAGACCCUUUAACCUGUGAGGACAUCCAGGGUCACAGGUGAGGUUCUUGGGAGCCUGGCGUCUGGCC
          Chromosome Location Chr. 19 - 51693254 - 51693339 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004602
          miRBase ID: hsa-miR-125a-3p
          Mature miRNA Sequence: ACAGGUGAGGUUCUUGGGAGCC
          Chromosome Location: Chr. 19 - 51693254 - 51693339 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-125a
          Stem-loop Accession # MI0000151
          Stem-loop Sequence
          CUGGGUCCCUGAGACCCUUUAACCUGUGAGGACGUCCAGGGUCACAGGUGAGGUUCUUGGGAGCCUGG
          Chromosome Location Chr. 17 - 17830812 - 17830879 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004528
          miRBase ID: mmu-miR-125a-3p
          Mature miRNA Sequence: ACAGGUGAGGUUCUUGGGAGCC
          Chromosome Location: Chr. 17 - 17830812 - 17830879 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-125a
          Stem-loop Accession # MI0000895
          Stem-loop Sequence
          UGCCGGCCUCUGGGUCCCUGAGACCCUUUAACCUGUGAGGACGUCCAGGGUCACAGGUGAGGUUCUUGGGAGCCUGGCGCCUGGC
          Chromosome Location Chr. 1 - 59704827 - 59704911 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004729
          miRBase ID: rno-miR-125a-3p
          Mature miRNA Sequence: ACAGGUGAGGUUCUUGGGAGCC
          Chromosome Location: Chr. 1 - 59704827 - 59704911 [+] on Build Rnor_6.0

          Guinea pig

          Stem-loop ID cpo-mir-125a
          Stem-loop Accession # MI0038624
          Stem-loop Sequence
          UCCCUGAGACCCUUUAACCUGUGAGGACAUCCAGGGUCACAGGUGAGGUUCUUGGGAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046966
          miRBase ID: cpo-miR-125a-3p
          Mature miRNA Sequence: ACAGGUGAGGUUCUUGGGAGCC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-125a
          Stem-loop Accession # MI0012760
          Stem-loop Sequence
          CUGGGUCCCUGAGACCCUUUAACCUGUGAGGACAUCCAGGGUCACAGGUGAGGUUCUUGGGAGCCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013009
          miRBase ID: eca-miR-125a-3p
          Mature miRNA Sequence: ACAGGUGAGGUUCUUGGGAGCC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-125a
          Stem-loop Accession # MI0008464
          Stem-loop Sequence
          GCCAGUCUCUAGGUCCCUGAGACCCUUUAACCUGUGAGGACAUCCAGGGUCACAGGUGAGGUUCUUGGGAGCCUGGUGUCUGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007984
          miRBase ID: ptr-miR-125a
          Mature miRNA Sequence: ACAGGUGAGGUUCUUGGGAGCC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-125a
          Stem-loop Accession # MI0014817
          Stem-loop Sequence
          UGCCAGUCUCUAGGUCCCUGAGACCCUUUAACCUGUGAGGACAUCCAGGGUCACAGGUGAGGUUCUUGGGAGCCUGGCGUCUGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015749
          miRBase ID: ppy-miR-125a-3p
          Mature miRNA Sequence: ACAGGUGAGGUUCUUGGGAGCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-125a
          Stem-loop Accession # MI0007615
          Stem-loop Sequence
          UGCCAGUCUCUGGGUCCCUGAGACCCUUUAACCUGUGAGGACAUCCAGGGUCACAGGUGAGGUUCUUGGGAGCCUGGCGUCUGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006182
          miRBase ID: mml-miR-125a-3p
          Mature miRNA Sequence: ACAGGUGAGGUUCUUGGGAGCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465947_pri
          TaqMan™ Pri-miRNA Assay : Mm03306233_pri
          TaqMan™ Pri-miRNA Assay : Hs03302570_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12378
          Ambion® Pre-miR™ miRNA Precursor : PM12378
          TaqMan™ MicroRNA Assay : 002199
          mirVana® miRNA inhibitor : MH12378
          mirVana® miRNA mimic : MC12378
          TaqMan™ Advanced miRNA Assay : 477883_mir
          TaqMan™ Advanced miRNA Assay : mmu481497_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline