Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM14757
          Assay Name: rno-miR-301a-5p
          miRBase Accession Number: MI0000593
          miRBase Version: v22.1
          Chromosome Location: Chr.10: 74417746 - 74417845 [+] on Build Rnor_6.0
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUAC
          Species: Rat, Goat, Ornithorhynchus anatinus
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM14757
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000034, mir-130

          Mature miRNA Sequence:

          GCUCUGACUUUAUUGCACUAC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-301a-5p MIMAT0017026
          Goat chi-miR-301a-5p MIMAT0036118
          Ornithorhynchus anatinus oan-miR-301-5p MIMAT0006909

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-301a
          Stem-loop Accession # MI0000593
          Stem-loop Sequence
          CCUGCUGGCUACUGCUGACGACUGCUCUGACUUUAUUGCACUACUGUACUGUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCCGGGAGCAGGCUAC
          Chromosome Location Chr. 10 - 74417746 - 74417845 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017026
          miRBase ID: rno-miR-301a-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUAC
          Chromosome Location: Chr. 10 - 74417746 - 74417845 [+] on Build Rnor_6.0

          Goat

          Stem-loop ID chi-mir-301a
          Stem-loop Accession # MI0030729
          Stem-loop Sequence
          ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036118
          miRBase ID: chi-miR-301a-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUAC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-301
          Stem-loop Accession # MI0006727
          Stem-loop Sequence
          UAUUAUGGUGUGUUUUACGCUUUCUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUUCUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGAAUACACAGCAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006909
          miRBase ID: oan-miR-301-5p
          Mature miRNA Sequence: GCUCUGACUUUAUUGCACUAC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464947_pri
          Ambion® Pre-miR™ miRNA Precursor : PM14757
          TaqMan™ MicroRNA Assay : 006497_mat
          mirVana® miRNA inhibitor : MH14757
          mirVana® miRNA mimic : MC14757
          TaqMan™ Advanced miRNA Assay : rno480881_mir


          miRBase Alias:

          Rat: rno-miR-301a* (v18)
          Ochrobactrum anthropi: oan-miR-301* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline