Search
Search

| Assay Name: | rno-miR-301a-5p |
|---|---|
| Stem-loop Accession Number: | MI0000593 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCUCUGACUUUAUUGCACUAC |
| Species: | Rat, Goat, Ornithorhynchus anatinus |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | GCUCUGACUUUAUUGCACUAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-301a-5p | MIMAT0017026 |
| Goat | chi-miR-301a-5p | MIMAT0036118 |
| Ornithorhynchus anatinus | oan-miR-301-5p | MIMAT0006909 |