Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM10211
          Assay Name: hsa-let-7i-5p
          miRBase Accession Number: MI0000434
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 62603686 - 62603769 [+] on Build GRCh38
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Species: Human, Mouse, Rat, Anolis carolinensis, Astatotilapia burtoni, Bovine, Callithrix jacchus, Chrysemys picta, Dasypus novemcinctus, Fugu rubripes, Goat, Guinea pig, Ictalurus punctatus, Metriaclima zebra, Neolamprologus brichardi, Nomascus leucogenys, Ophiophagus hannah, Oreochromis niloticus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pongo pygmaeus, Pteropus alecto, Pundamilia nyererei, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Sheep, Taeniopygia guttata, Tetraodon nigroviridis, Tupaia chinensis, Zebrafish
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM10211
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          UGAGGUAGUAGUUUGUGCUGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7i-5p MIMAT0000415
          Mouse mmu-let-7i-5p MIMAT0000122
          Rat rno-let-7i-5p MIMAT0000779
          Anolis carolinensis aca-let-7i-5p MIMAT0021707
          Astatotilapia burtoni abu-let-7i MIMAT0042087
          Bovine bta-let-7i MIMAT0003851
          View More Rows
          Callithrix jacchus cja-let-7i MIMAT0049305
          Chrysemys picta cpi-let-7i-5p MIMAT0037610
          Dasypus novemcinctus dno-let-7i-5p MIMAT0047486
          Fugu rubripes fru-let-7i MIMAT0002977
          Goat chi-let-7i-5p MIMAT0035894
          Guinea pig cpo-let-7i-5p MIMAT0046833
          Ictalurus punctatus ipu-let-7i MIMAT0029382
          Metriaclima zebra mze-let-7i MIMAT0042312
          Neolamprologus brichardi nbr-let-7i MIMAT0042515
          Nomascus leucogenys nle-let-7i MIMAT0049470
          Ophiophagus hannah oha-let-7i-5p MIMAT0036642
          Oreochromis niloticus oni-let-7i MIMAT0042682
          Otolemur garnettii oga-let-7i MIMAT0049705
          Pan paniscus ppa-let-7i MIMAT0049063
          Pan troglodytes ptr-let-7i MIMAT0007943
          Papio hamadryas pha-let-7i MIMAT0049571
          Pongo pygmaeus ppy-let-7i MIMAT0015728
          Pteropus alecto pal-let-7i-5p MIMAT0039786
          Pundamilia nyererei pny-let-7i MIMAT0042898
          Python bivittatus pbv-let-7i-5p MIMAT0038810
          Rabbit ocu-let-7i-5p MIMAT0048089
          Rhesus monkey mml-let-7i-5p MIMAT0006158
          Salmo salar ssa-let-7i-5p MIMAT0032274
          Sheep oar-let-7i MIMAT0030026
          Taeniopygia guttata tgu-let-7i-5p MIMAT0014504
          Tetraodon nigroviridis tni-let-7i MIMAT0002978
          Tupaia chinensis tch-let-7i-5p MIMAT0036604
          Zebrafish dre-let-7i MIMAT0001767

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7i
          Stem-loop Accession # MI0000434
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUA
          Chromosome Location Chr. 12 - 62603686 - 62603769 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000415
          miRBase ID: hsa-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: Chr. 12 - 62603686 - 62603769 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-let-7i
          Stem-loop Accession # MI0000138
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAG
          Chromosome Location Chr. 10 - 122985640 - 122985724 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000122
          miRBase ID: mmu-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: Chr. 10 - 122985640 - 122985724 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-let-7i
          Stem-loop Accession # MI0000835
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAG
          Chromosome Location Chr. 7 - 66802731 - 66802815 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000779
          miRBase ID: rno-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: Chr. 7 - 66802731 - 66802815 [+] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-let-7i
          Stem-loop Accession # MI0018710
          Stem-loop Sequence
          GCACCACGGCCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGCUGGUGAUGAUCGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021707
          miRBase ID: aca-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Astatotilapia burtoni

          Stem-loop ID abu-let-7i
          Stem-loop Accession # MI0033784
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUUGGGUUGAGACACUGCCCGCUAUGGAGAUGACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042087
          miRBase ID: abu-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-let-7i
          Stem-loop Accession # MI0005065
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003851
          miRBase ID: bta-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-let-7i
          Stem-loop Accession # MI0040041
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049305
          miRBase ID: cja-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-let-7i
          Stem-loop Accession # MI0029383
          Stem-loop Sequence
          CCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037610
          miRBase ID: cpi-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-let-7i
          Stem-loop Accession # MI0038894
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047486
          miRBase ID: dno-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-let-7i
          Stem-loop Accession # MI0003295
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUUGGGUUGUGACACUGCCCGCUAUGGAGAUGACUGCGCAAGCUACUGCCUUGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002977
          miRBase ID: fru-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-let-7i
          Stem-loop Accession # MI0030586
          Stem-loop Sequence
          CCCGACACCAUGGCCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGCUGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035894
          miRBase ID: chi-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7i
          Stem-loop Accession # MI0038550
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046833
          miRBase ID: cpo-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-let-7i
          Stem-loop Accession # MI0024452
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUUGGGAUGUGACAUUGCCCGUUAUGGAGAUGACUGCGCAAGCUACUCCCUUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029382
          miRBase ID: ipu-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-let-7i-1
          Stem-loop Accession # MI0034075
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUUGGGUUGAGACACUGCCCGCUAUGGAGAUGACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042312
          miRBase ID: mze-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Stem-loop ID mze-let-7i-2
          Stem-loop Accession # MI0034076
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUCAUAUUACCCCUUGCGGAGAUGACUGCGCAAUCUACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042312
          miRBase ID: mze-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-let-7i-1
          Stem-loop Accession # MI0034354
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUUGGGUUGAGACACUGCCCGCUAUGGAGAUGACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042515
          miRBase ID: nbr-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Stem-loop ID nbr-let-7i-2
          Stem-loop Accession # MI0034355
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUCAUAUUACCCCUUGCGGAGAUGACUGCGCAAUCUACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042515
          miRBase ID: nbr-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-let-7i
          Stem-loop Accession # MI0040226
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049470
          miRBase ID: nle-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-let-7i
          Stem-loop Accession # MI0031310
          Stem-loop Sequence
          UGACCCUGGCUGAGGUAGUAGUUUGUGCUGUUCGUCGGGCUGUGAUACUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036642
          miRBase ID: oha-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-let-7i-1
          Stem-loop Accession # MI0034588
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUUGGGUUGAGACACUGCCCGCUAUGGAGAUGACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042682
          miRBase ID: oni-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Stem-loop ID oni-let-7i-2
          Stem-loop Accession # MI0034589
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUCAUAUUACCCCUUGUGGAGAUGACUGCGCAAUCUACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042682
          miRBase ID: oni-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-let-7i
          Stem-loop Accession # MI0040489
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGCUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049705
          miRBase ID: oga-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-let-7i
          Stem-loop Accession # MI0039771
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049063
          miRBase ID: ppa-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-let-7i
          Stem-loop Accession # MI0008407
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007943
          miRBase ID: ptr-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-let-7i
          Stem-loop Accession # MI0040342
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049571
          miRBase ID: pha-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-let-7i
          Stem-loop Accession # MI0014789
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015728
          miRBase ID: ppy-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-let-7i
          Stem-loop Accession # MI0032327
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039786
          miRBase ID: pal-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-let-7i-1
          Stem-loop Accession # MI0034867
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUUGGGUUGAGACACUGCCCGCUAUGGAGAUGACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042898
          miRBase ID: pny-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Stem-loop ID pny-let-7i-2
          Stem-loop Accession # MI0034868
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUCAUAUUACCCCUUGCGGAGAUGACUGCGCAAUCUACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042898
          miRBase ID: pny-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-let-7i
          Stem-loop Accession # MI0030142
          Stem-loop Sequence
          ACCCUGGCUGAGGUAGUAGUUUGUGCUGUUCGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038810
          miRBase ID: pbv-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-let-7i
          Stem-loop Accession # MI0039214
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048089
          miRBase ID: ocu-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-let-7i
          Stem-loop Accession # MI0007580
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006158
          miRBase ID: mml-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-let-7i-1
          Stem-loop Accession # MI0026429
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUGGCCUGCUGUGGAGAUAACUGCGCAAACGACUACCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032274
          miRBase ID: ssa-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Stem-loop ID ssa-let-7i-2
          Stem-loop Accession # MI0026430
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUUGGGUUAUGACAUUUCCCGCUAUGGAGAUGACUGCGCAAGCUACUGCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032274
          miRBase ID: ssa-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-let-7i
          Stem-loop Accession # MI0025247
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030026
          miRBase ID: oar-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-let-7i
          Stem-loop Accession # MI0013694
          Stem-loop Sequence
          GGGCCCUGGCUGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCUAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014504
          miRBase ID: tgu-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-let-7i
          Stem-loop Accession # MI0003296
          Stem-loop Sequence
          CUGGCUGAGGUAGUAGUUUGUGCUGUUGGUUGGGUUGUGACACUGCCCGCUAUGGAGAUGACUGCGCAAGCUACUGCCUUGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002978
          miRBase ID: tni-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-let-7i
          Stem-loop Accession # MI0031272
          Stem-loop Sequence
          UGAGGUAGUAGUUUGUGCUGUUGGUCGGGUUGUGACAUUGCCCGCUGUGGAGAUAACUGCGCAAGCUACUGCCUUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036604
          miRBase ID: tch-let-7i-5p
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-let-7i
          Stem-loop Accession # MI0001876
          Stem-loop Sequence
          GUGUACUGGCUGAGGUAGUAGUUUGUGCUGUUGGUUGGGAUGUGACAUUGCCCGUUAUGGAGAUGACUGCGCAAGCUACUCCCUUGCCAGUGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001767
          miRBase ID: dre-let-7i
          Mature miRNA Sequence: UGAGGUAGUAGUUUGUGCUGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306160_pri
          TaqMan™ Pri-miRNA Assay : Rn03465226_pri
          TaqMan™ Pri-miRNA Assay : Hs03303039_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10211
          TaqMan™ MicroRNA Assay : 002221
          mirVana® miRNA inhibitor : MH10211
          mirVana® miRNA mimic : MC10211
          TaqMan™ Advanced miRNA Assay : rno478375_mir
          TaqMan™ Advanced miRNA Assay : 478375_mir


          miRBase Alias:

          Human: hsa-let-7i (v17)
          Mouse: mmu-let-7i (v17)
          Rat: rno-let-7i (v18)
          Aeromonas caviae Ae398: aca-let-7i (v18)
          Macaca mulatta: mml-let-7i (v19)
          Trabulsiella guamensis ATCC 49490: tgu-let-7i (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline