Search
Search

| Assay Name: | hsa-let-7i |
|---|---|
| miRBase Accession Number: | MI0000434 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGGUAGUAGUUUGUGCUGUU |
| Species: | Human, Mouse, Rat, Anolis carolinensis, Astatotilapia burtoni, Bovine, Callithrix jacchus, Chrysemys picta, Dasypus novemcinctus, Fugu rubripes, Goat, Guinea pig, Ictalurus punctatus, Metriaclima zebra, Neolamprologus brichardi, Nomascus leucogenys, Ophiophagus hannah, Oreochromis niloticus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pongo pygmaeus, Pteropus alecto, Pundamilia nyererei, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Sheep, Taeniopygia guttata, Tetraodon nigroviridis, Tupaia chinensis, Zebrafish |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | UGAGGUAGUAGUUUGUGCUGUU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-let-7i-5p | MIMAT0000415 | |
| Mouse | mmu-let-7i-5p | MIMAT0000122 | |
| Rat | rno-let-7i-5p | MIMAT0000779 | |
| Anolis carolinensis | aca-let-7i-5p | MIMAT0021707 | |
| Astatotilapia burtoni | abu-let-7i | MIMAT0042087 | |
| Bovine | bta-let-7i | MIMAT0003851 | |
| View More Rows | |||
| Callithrix jacchus | cja-let-7i | MIMAT0049305 | |
| Chrysemys picta | cpi-let-7i-5p | MIMAT0037610 | |
| Dasypus novemcinctus | dno-let-7i-5p | MIMAT0047486 | |
| Fugu rubripes | fru-let-7i | MIMAT0002977 | |
| Goat | chi-let-7i-5p | MIMAT0035894 | |
| Guinea pig | cpo-let-7i-5p | MIMAT0046833 | |
| Ictalurus punctatus | ipu-let-7i | MIMAT0029382 | |
| Metriaclima zebra | mze-let-7i | MIMAT0042312 | |
| Neolamprologus brichardi | nbr-let-7i | MIMAT0042515 | |
| Nomascus leucogenys | nle-let-7i | MIMAT0049470 | |
| Ophiophagus hannah | oha-let-7i-5p | MIMAT0036642 | |
| Oreochromis niloticus | oni-let-7i | MIMAT0042682 | |
| Otolemur garnettii | oga-let-7i | MIMAT0049705 | |
| Pan paniscus | ppa-let-7i | MIMAT0049063 | |
| Pan troglodytes | ptr-let-7i | MIMAT0007943 | |
| Papio hamadryas | pha-let-7i | MIMAT0049571 | |
| Pongo pygmaeus | ppy-let-7i | MIMAT0015728 | |
| Pteropus alecto | pal-let-7i-5p | MIMAT0039786 | |
| Pundamilia nyererei | pny-let-7i | MIMAT0042898 | |
| Python bivittatus | pbv-let-7i-5p | MIMAT0038810 | |
| Rabbit | ocu-let-7i-5p | MIMAT0048089 | |
| Rhesus monkey | mml-let-7i-5p | MIMAT0006158 | |
| Salmo salar | ssa-let-7i-5p | MIMAT0032274 | |
| Sheep | oar-let-7i | MIMAT0030026 | |
| Taeniopygia guttata | tgu-let-7i-5p | MIMAT0014504 | |
| Tetraodon nigroviridis | tni-let-7i | MIMAT0002978 | |
| Tupaia chinensis | tch-let-7i-5p | MIMAT0036604 | |
| Zebrafish | dre-let-7i | MIMAT0001767 | |