Search
Search

| Assay Name: | hsa-miR-486-5p |
|---|---|
| miRBase Accession Number: | MI0002470 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCUGUACUGAGCUGCCCCGAG |
| Species: | Human, Mouse, Rat, Bovine, Cricetulus griseus, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000220, mir-486 |
Mature miRNA Sequence: | UCCUGUACUGAGCUGCCCCGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-486-5p | MIMAT0002177 |
| Mouse | mmu-miR-486a-5p | MIMAT0003130 |
| Mouse | mmu-miR-486b-5p | MIMAT0014943 |
| Rat | rno-miR-486 | MIMAT0037265 |
| Bovine | bta-miR-486 | MIMAT0009329 |
| Cricetulus griseus | cgr-miR-486-5p | MIMAT0023967 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-486-5p | MIMAT0047912 |
| Gorilla gorilla | ggo-miR-486 | MIMAT0024149 |
| Guinea pig | cpo-miR-486-5p | MIMAT0047277 |
| Horse | eca-miR-486-5p | MIMAT0013186 |
| Pan troglodytes | ptr-miR-486 | MIMAT0008160 |
| Pig | ssc-miR-486 | MIMAT0013886 |
| Pongo pygmaeus | ppy-miR-486-5p | MIMAT0015904 |
| Pteropus alecto | pal-miR-486a-5p | MIMAT0039846 |
| Rhesus monkey | mml-miR-486-5p | MIMAT0006344 |