Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10546
          Assay Name: hsa-miR-486-5p
          miRBase Accession Number: MI0002470
          miRBase Version: v22.1
          Chromosome Location: Chr.8: 41660441 - 41660508 [-] on Build GRCh38
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Species: Human, Mouse, Rat, Bovine, Cricetulus griseus, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10546
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000220, mir-486

          Mature miRNA Sequence:

          UCCUGUACUGAGCUGCCCCGAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-486-5p MIMAT0002177
          Mouse mmu-miR-486a-5p MIMAT0003130
          Mouse mmu-miR-486b-5p MIMAT0014943
          Rat rno-miR-486 MIMAT0037265
          Bovine bta-miR-486 MIMAT0009329
          Cricetulus griseus cgr-miR-486-5p MIMAT0023967
          View More Rows
          Dasypus novemcinctus dno-miR-486-5p MIMAT0047912
          Gorilla gorilla ggo-miR-486 MIMAT0024149
          Guinea pig cpo-miR-486-5p MIMAT0047277
          Horse eca-miR-486-5p MIMAT0013186
          Pan troglodytes ptr-miR-486 MIMAT0008160
          Pig ssc-miR-486 MIMAT0013886
          Pongo pygmaeus ppy-miR-486-5p MIMAT0015904
          Pteropus alecto pal-miR-486a-5p MIMAT0039846
          Rhesus monkey mml-miR-486-5p MIMAT0006344

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-486-1
          Stem-loop Accession # MI0002470
          Stem-loop Sequence
          GCAUCCUGUACUGAGCUGCCCCGAGGCCCUUCAUGCUGCCCAGCUCGGGGCAGCUCAGUACAGGAUAC
          Chromosome Location Chr. 8 - 41660441 - 41660508 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002177
          miRBase ID: hsa-miR-486-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: Chr. 8 - 41660441 - 41660508 [-] on Build GRCh38

          Stem-loop ID hsa-mir-486-2
          Stem-loop Accession # MI0023622
          Stem-loop Sequence
          UCCUGUACUGAGCUGCCCCGAGCUGGGCAGCAUGAAGGGCCUCGGGGCAGCUCAGUACAGGAUG
          Chromosome Location Chr. 8 - 41660444 - 41660507 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002177
          miRBase ID: hsa-miR-486-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: Chr. 8 - 41660441 - 41660508 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-486a
          Stem-loop Accession # MI0003493
          Stem-loop Sequence
          CAGCCAGCUCUGAUCUCGCCCUCCCUGAGGGGUCCUGUACUGAGCUGCCCCGAGGUCCUUCACUGUGCUCAGCUCGGGGCAGCUCAGUACAGGAUGCGUCAGGGUGGGAGACAACGGGGAACAAGCCA
          Chromosome Location Chr. 8 - 23142555 - 23142682 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003130
          miRBase ID: mmu-miR-486a-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: Chr. 8 - 23142555 - 23142682 [+] on Build GRCm38

          Stem-loop ID mmu-mir-486b
          Stem-loop Accession # MI0014103
          Stem-loop Sequence
          CCCACCCUGACGCAUCCUGUACUGAGCUGCCCCGAGCUGAGCACAGUGAAGGACCUCGGGGCAGCUCAGUACAGGACCCCUCAGGGAGGG
          Chromosome Location Chr. 8 - 23142573 - 23142662 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0014943
          miRBase ID: mmu-miR-486b-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: Chr. 8 - 23142573 - 23142662 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-486
          Stem-loop Accession # MI0031787
          Stem-loop Sequence
          GCAUCCUGUACUGAGCUGCCCCGAGCUGAGCACAAUGAAGGACCUCGGGGCAGCUCAGUACAGGACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037265
          miRBase ID: rno-miR-486
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-486
          Stem-loop Accession # MI0009843
          Stem-loop Sequence
          GCCAGCUUGGACCUGCGUCCUCCCUGACGGGUCCUGUACUGAGCUGCCCCGAGGCCCUUCGCUGUGCCCAGCUCGGGUCAGCUCAGUACCGGGCGCGUCGGGGUGGGAGUCGGCCGGAAGCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009329
          miRBase ID: bta-miR-486
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-486
          Stem-loop Accession # MI0020530
          Stem-loop Sequence
          CCUGAGGGGUCCUGUACUGAGCUGCCCCGAGGUCCUUCAUUGUGCUCAGCUCGGGGCAGCUCAGUACAAGACGCAUCGGGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023967
          miRBase ID: cgr-miR-486-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-486
          Stem-loop Accession # MI0039122
          Stem-loop Sequence
          UCCUGUACUGAGCUGCCCCGAGGCCCUGCCUUGUGCCCCACUCGGGGCAGCUCAGUACAGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047912
          miRBase ID: dno-miR-486-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-486
          Stem-loop Accession # MI0020696
          Stem-loop Sequence
          UCUCCAUCCUCCCUGGGGCAUCCUGUACUGAGCUGCCCCGAGGCCCUUCAUGCUGCCCAGCUCGGGGCAGCUCAGUACAGGAUACCUCGGGGUGGGAGUCAGCAGGAGGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024149
          miRBase ID: ggo-miR-486
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-486
          Stem-loop Accession # MI0038788
          Stem-loop Sequence
          UCCUGUACUGAGCUGCCCCGAGGCCCUUCCUUGUGCCCCGCUCGGGGCAGCUCAGUACAGGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047277
          miRBase ID: cpo-miR-486-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-486
          Stem-loop Accession # MI0012931
          Stem-loop Sequence
          GGAUCCUGUACUGAGCUGCCCCGAGGCCCUUCACUGUGCCCAGCUCGGGGCAGCUCAGUACAGGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013186
          miRBase ID: eca-miR-486-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-486
          Stem-loop Accession # MI0008682
          Stem-loop Sequence
          GCAUCCUGUACUGAGCUGCCCCGAGGCCCUUCAUGCUGCCCAGCUCGGGGCAGCUCAGUACAGGAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008160
          miRBase ID: ptr-miR-486
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-486-1
          Stem-loop Accession # MI0013103
          Stem-loop Sequence
          GCUGGGGCUUCAGGGCCAGCUCGGACCUCUGACCUCCCUGACGGGUCCUGUACUGAGCUGCCCCGAGGCCCUCACUGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013886
          miRBase ID: ssc-miR-486
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Stem-loop ID ssc-mir-486-2
          Stem-loop Accession # MI0017992
          Stem-loop Sequence
          CCCUGACGGGUCCUGUACUGAGCUGCCCCGAGGCCCUCACUGUGCCCAGCUCGGGCCAGCUCAGUACCCGGCGCGUCGGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013886
          miRBase ID: ssc-miR-486
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-486
          Stem-loop Accession # MI0014958
          Stem-loop Sequence
          GCAUCCUGUACUGAGCUGCCCCGAGGCCCUUCAUGCUGCCCAGCUCGGGGCAGCUCAGUACAGGAUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015904
          miRBase ID: ppy-miR-486-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-486a-1
          Stem-loop Accession # MI0032359
          Stem-loop Sequence
          UCCUGUACUGAGCUGCCCCGAGGCUCUUCACUGUGCCCAGCUCGGGGCAGCUCAGUACAGGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039846
          miRBase ID: pal-miR-486a-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Stem-loop ID pal-mir-486a-2
          Stem-loop Accession # MI0032361
          Stem-loop Sequence
          UCCUGUACUGAGCUGCCCCGAGCUGGGCACAGUGAAGAGCCUCGGGGCAGCUCAGUACAGGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039846
          miRBase ID: pal-miR-486a-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Stem-loop ID pal-mir-486a-3
          Stem-loop Accession # MI0032589
          Stem-loop Sequence
          UCCUGUACUGAGCUGCCCCGAGGCUCUUCACUGUGCCCAGCUCGGGGCAGCUGAGCAUGGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039846
          miRBase ID: pal-miR-486a-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-486
          Stem-loop Accession # MI0007759
          Stem-loop Sequence
          GUAUCCUGUACUGAGCUGCCCCGAGCUGGGCAGCAUGAAGGGCCUCGGGGCAGCUCAGUACAGGAUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006344
          miRBase ID: mml-miR-486-5p
          Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303815_pri
          TaqMan™ Pri-miRNA Assay : Mm03307639_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10546
          TaqMan™ MicroRNA Assay : 001278
          mirVana® miRNA inhibitor : MH10546
          mirVana® miRNA mimic : MC10546
          TaqMan™ Advanced miRNA Assay : rno481183_mir
          TaqMan™ Advanced miRNA Assay : 478128_mir
          TaqMan™ Advanced miRNA Assay : mmu481183_mir


          miRBase Alias:

          Human: hsa-miR-486 (v9.2)
          Mouse: mmu-miR-486-5p (v20)
          Mouse: mmu-miR-3107-5p (v20)
          Mouse: mmu-miR-3107 (v17)
          Mouse: mmu-miR-486 (v17)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline