Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM10774
          Assay Name: hsa-miR-433-3p
          miRBase Accession Number: MI0001723
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100881886 - 100881978 [+] on Build GRCh38
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Species: Human, Mouse, Rat, Bovine, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Sheep
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM10774
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000177, mir-433

          Mature miRNA Sequence:

          AUCAUGAUGGGCUCCUCGGUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-433-3p MIMAT0001627
          Mouse mmu-miR-433-3p MIMAT0001420
          Rat rno-miR-433-3p MIMAT0001628
          Bovine bta-miR-433 MIMAT0009318
          Dasypus novemcinctus dno-miR-433-3p MIMAT0047891
          Dog cfa-miR-433 MIMAT0006712
          View More Rows
          Goat chi-miR-433 MIMAT0036229
          Guinea pig cpo-miR-433-3p MIMAT0047254
          Horse eca-miR-433 MIMAT0013158
          Pan troglodytes ptr-miR-433 MIMAT0008147
          Pongo pygmaeus ppy-miR-433 MIMAT0015889
          Pteropus alecto pal-miR-433-3p MIMAT0040299
          Rabbit ocu-miR-433-3p MIMAT0048450
          Rhesus monkey mml-miR-433-3p MIMAT0006328
          Sheep oar-miR-433-3p MIMAT0019242

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-433
          Stem-loop Accession # MI0001723
          Stem-loop Sequence
          CCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location Chr. 14 - 100881886 - 100881978 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001627
          miRBase ID: hsa-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: Chr. 14 - 100881886 - 100881978 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-433
          Stem-loop Accession # MI0001525
          Stem-loop Sequence
          UGCCCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGGUAGCGGCACCACACCAUGAAGGCAGCCC
          Chromosome Location Chr. 12 - 109591715 - 109591838 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001420
          miRBase ID: mmu-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: Chr. 12 - 109591715 - 109591838 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-433
          Stem-loop Accession # MI0001724
          Stem-loop Sequence
          CCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location Chr. 6 - 133712334 - 133712426 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0001628
          miRBase ID: rno-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: Chr. 6 - 133712334 - 133712426 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-433
          Stem-loop Accession # MI0009832
          Stem-loop Sequence
          UGGCCGGGGAGAAGUACGGUGAGCCUGUCAUUGUUCAGAGAGGCUAGAUCCUCCUUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGGUAGCGGCACCACACCAUGAAGGCAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009318
          miRBase ID: bta-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-433
          Stem-loop Accession # MI0039110
          Stem-loop Sequence
          UACGGUGAGCCUGUCAUUAUUCAGAGAGGCAUGAUCCUCAUCUUUGAGAAGAAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047891
          miRBase ID: dno-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-433
          Stem-loop Accession # MI0008131
          Stem-loop Sequence
          UACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006712
          miRBase ID: cfa-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-433
          Stem-loop Accession # MI0030793
          Stem-loop Sequence
          ACCCAGUGGCCGGGGAGAAGUACGGUGAGCCUGUCAUUGUUCAGAGAGGCUAGAUCCUCCUUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGGUAGCGGCACCACACCAUGAAGGCAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036229
          miRBase ID: chi-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-433
          Stem-loop Accession # MI0038775
          Stem-loop Sequence
          UACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047254
          miRBase ID: cpo-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-433
          Stem-loop Accession # MI0012902
          Stem-loop Sequence
          UGCCCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUAAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGGUAGCGGCACCACACCAUGAAGGCAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013158
          miRBase ID: eca-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-433
          Stem-loop Accession # MI0008668
          Stem-loop Sequence
          CGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008147
          miRBase ID: ptr-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-433
          Stem-loop Accession # MI0014945
          Stem-loop Sequence
          CCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015889
          miRBase ID: ppy-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-433
          Stem-loop Accession # MI0032599
          Stem-loop Sequence
          UGCGGUGAGCCUGUCAUCGUUCAGAGAGGCCAGGUCCUCCUUGUCCAGAAGGAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040299
          miRBase ID: pal-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-433
          Stem-loop Accession # MI0039406
          Stem-loop Sequence
          UACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCGCUGCUGAGAAGGAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048450
          miRBase ID: ocu-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-433
          Stem-loop Accession # MI0007745
          Stem-loop Sequence
          CCAGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006328
          miRBase ID: mml-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-433
          Stem-loop Accession # MI0016914
          Stem-loop Sequence
          ACCCAGUGGCCGGGGAGAAGUACGGUGAGCCUGUCAUUGUUCAGAGAGGCUAGAUCCUCCUUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGGUAGCGGCACCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019242
          miRBase ID: oar-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303744_pri
          TaqMan™ Pri-miRNA Assay : Rn03466640_pri
          TaqMan™ Pri-miRNA Assay : Mm03307482_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10774
          TaqMan™ MicroRNA Assay : 001028
          mirVana® miRNA inhibitor : MH10774
          mirVana® miRNA mimic : MC10774
          TaqMan™ Advanced miRNA Assay : mmu481838_mir
          TaqMan™ Advanced miRNA Assay : 478102_mir
          TaqMan™ Advanced miRNA Assay : rno481164_mir


          miRBase Alias:

          Human: hsa-miR-433 (v19)
          Mouse: mmu-miR-433 (v17)
          Rat: rno-miR-433 (v18)
          Macaca mulatta: mml-miR-433 (v19)

          Back To Top

          Related Products

          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline