Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10774
          Assay Name: hsa-miR-433-3p
          miRBase Accession Number: MI0001723
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100881886 - 100881978 [+] on Build GRCh38
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Species: Human, Mouse, Rat, Bovine, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Sheep
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10774

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000177, mir-433

          Mature miRNA Sequence:

          AUCAUGAUGGGCUCCUCGGUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-433-3p MIMAT0001627
          Mouse mmu-miR-433-3p MIMAT0001420
          Rat rno-miR-433-3p MIMAT0001628
          Bovine bta-miR-433 MIMAT0009318
          Dasypus novemcinctus dno-miR-433-3p MIMAT0047891
          Dog cfa-miR-433 MIMAT0006712
          View More Rows
          Goat chi-miR-433 MIMAT0036229
          Guinea pig cpo-miR-433-3p MIMAT0047254
          Horse eca-miR-433 MIMAT0013158
          Pan troglodytes ptr-miR-433 MIMAT0008147
          Pongo pygmaeus ppy-miR-433 MIMAT0015889
          Pteropus alecto pal-miR-433-3p MIMAT0040299
          Rabbit ocu-miR-433-3p MIMAT0048450
          Rhesus monkey mml-miR-433-3p MIMAT0006328
          Sheep oar-miR-433-3p MIMAT0019242

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-433
          Stem-loop Accession # MI0001723
          Stem-loop Sequence
          CCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location Chr. 14 - 100881886 - 100881978 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001627
          miRBase ID: hsa-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: Chr. 14 - 100881886 - 100881978 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-433
          Stem-loop Accession # MI0001525
          Stem-loop Sequence
          UGCCCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGGUAGCGGCACCACACCAUGAAGGCAGCCC
          Chromosome Location Chr. 12 - 109591715 - 109591838 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001420
          miRBase ID: mmu-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: Chr. 12 - 109591715 - 109591838 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-433
          Stem-loop Accession # MI0001724
          Stem-loop Sequence
          CCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location Chr. 6 - 133712334 - 133712426 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0001628
          miRBase ID: rno-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: Chr. 6 - 133712334 - 133712426 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-433
          Stem-loop Accession # MI0009832
          Stem-loop Sequence
          UGGCCGGGGAGAAGUACGGUGAGCCUGUCAUUGUUCAGAGAGGCUAGAUCCUCCUUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGGUAGCGGCACCACACCAUGAAGGCAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009318
          miRBase ID: bta-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-433
          Stem-loop Accession # MI0039110
          Stem-loop Sequence
          UACGGUGAGCCUGUCAUUAUUCAGAGAGGCAUGAUCCUCAUCUUUGAGAAGAAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047891
          miRBase ID: dno-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-433
          Stem-loop Accession # MI0008131
          Stem-loop Sequence
          UACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006712
          miRBase ID: cfa-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-433
          Stem-loop Accession # MI0030793
          Stem-loop Sequence
          ACCCAGUGGCCGGGGAGAAGUACGGUGAGCCUGUCAUUGUUCAGAGAGGCUAGAUCCUCCUUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGGUAGCGGCACCACACCAUGAAGGCAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036229
          miRBase ID: chi-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-433
          Stem-loop Accession # MI0038775
          Stem-loop Sequence
          UACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047254
          miRBase ID: cpo-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-433
          Stem-loop Accession # MI0012902
          Stem-loop Sequence
          UGCCCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUAAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGGUAGCGGCACCACACCAUGAAGGCAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013158
          miRBase ID: eca-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-433
          Stem-loop Accession # MI0008668
          Stem-loop Sequence
          CGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008147
          miRBase ID: ptr-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-433
          Stem-loop Accession # MI0014945
          Stem-loop Sequence
          CCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015889
          miRBase ID: ppy-miR-433
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-433
          Stem-loop Accession # MI0032599
          Stem-loop Sequence
          UGCGGUGAGCCUGUCAUCGUUCAGAGAGGCCAGGUCCUCCUUGUCCAGAAGGAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040299
          miRBase ID: pal-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-433
          Stem-loop Accession # MI0039406
          Stem-loop Sequence
          UACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCGCUGCUGAGAAGGAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048450
          miRBase ID: ocu-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-433
          Stem-loop Accession # MI0007745
          Stem-loop Sequence
          CCAGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006328
          miRBase ID: mml-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-433
          Stem-loop Accession # MI0016914
          Stem-loop Sequence
          ACCCAGUGGCCGGGGAGAAGUACGGUGAGCCUGUCAUUGUUCAGAGAGGCUAGAUCCUCCUUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGGUAGCGGCACCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019242
          miRBase ID: oar-miR-433-3p
          Mature miRNA Sequence: AUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303744_pri
          TaqMan™ Pri-miRNA Assay : Rn03466640_pri
          TaqMan™ Pri-miRNA Assay : Mm03307482_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10774
          Ambion® Pre-miR™ miRNA Precursor : PM10774
          TaqMan™ MicroRNA Assay : 001028
          mirVana® miRNA mimic : MC10774
          TaqMan™ Advanced miRNA Assay : mmu481838_mir
          TaqMan™ Advanced miRNA Assay : 478102_mir
          TaqMan™ Advanced miRNA Assay : rno481164_mir


          miRBase Alias:

          Human: hsa-miR-433 (v19)
          Mouse: mmu-miR-433 (v17)
          Rat: rno-miR-433 (v18)
          Macaca mulatta: mml-miR-433 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline