Search
Search

| Assay Name: | hsa-miR-215-5p |
|---|---|
| miRBase Accession Number: | MI0000291 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUGACCUAUGAAUUGACAGAC |
| Species: | Human, Alligator mississippiensis, Chicken, Chrysemys picta, Goat, Gorilla gorilla, Horse, Macaca nemestrina, Ornithorhynchus anatinus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey, Taeniopygia guttata |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000063, mir-192 |
Mature miRNA Sequence: | AUGACCUAUGAAUUGACAGAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-215-5p | MIMAT0000272 |
| Alligator mississippiensis | ami-miR-215-5p | MIMAT0038240 |
| Chicken | gga-miR-215-5p | MIMAT0001134 |
| Chrysemys picta | cpi-miR-215-5p | MIMAT0037841 |
| Goat | chi-miR-215-5p | MIMAT0036060 |
| Gorilla gorilla | ggo-miR-215 | MIMAT0002734 |
| View More Rows | ||
| Horse | eca-miR-215 | MIMAT0013190 |
| Macaca nemestrina | mne-miR-215 | MIMAT0002736 |
| Ornithorhynchus anatinus | oan-miR-215-5p | MIMAT0007000 |
| Pan troglodytes | ptr-miR-215 | MIMAT0002730 |
| Pig | ssc-miR-215 | MIMAT0010192 |
| Pongo pygmaeus | ppy-miR-215 | MIMAT0002732 |
| Rhesus monkey | mml-miR-215-5p | MIMAT0002728 |
| Taeniopygia guttata | tgu-miR-215-5p | MIMAT0014577 |