Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10874
          Assay Name: hsa-miR-215-5p
          miRBase Accession Number: MI0000291
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 220117853 - 220117962 [-] on Build GRCh38
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Species: Human, Alligator mississippiensis, Chicken, Chrysemys picta, Goat, Gorilla gorilla, Horse, Macaca nemestrina, Ornithorhynchus anatinus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey, Taeniopygia guttata
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10874

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000063, mir-192

          Mature miRNA Sequence:

          AUGACCUAUGAAUUGACAGAC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-215-5p MIMAT0000272
          Alligator mississippiensis ami-miR-215-5p MIMAT0038240
          Chicken gga-miR-215-5p MIMAT0001134
          Chrysemys picta cpi-miR-215-5p MIMAT0037841
          Goat chi-miR-215-5p MIMAT0036060
          Gorilla gorilla ggo-miR-215 MIMAT0002734
          View More Rows
          Horse eca-miR-215 MIMAT0013190
          Macaca nemestrina mne-miR-215 MIMAT0002736
          Ornithorhynchus anatinus oan-miR-215-5p MIMAT0007000
          Pan troglodytes ptr-miR-215 MIMAT0002730
          Pig ssc-miR-215 MIMAT0010192
          Pongo pygmaeus ppy-miR-215 MIMAT0002732
          Rhesus monkey mml-miR-215-5p MIMAT0002728
          Taeniopygia guttata tgu-miR-215-5p MIMAT0014577

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-215
          Stem-loop Accession # MI0000291
          Stem-loop Sequence
          AUCAUUCAGAAAUGGUAUACAGGAAAAUGACCUAUGAAUUGACAGACAAUAUAGCUGAGUUUGUCUGUCAUUUCUUUAGGCCAAUAUUCUGUAUGACUGUGCUACUUCAA
          Chromosome Location Chr. 1 - 220117853 - 220117962 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000272
          miRBase ID: hsa-miR-215-5p
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: Chr. 1 - 220117853 - 220117962 [-] on Build GRCh38

          Alligator mississippiensis

          Stem-loop ID ami-mir-215
          Stem-loop Accession # MI0029798
          Stem-loop Sequence
          AACUGUUGUGCAGGAAAAUGACCUAUGAAUUGACAGACUGUGUAUUCUAAGCUUGUCUGUCAUUUCUGUAGGCCAAUAUUCUGCACACCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038240
          miRBase ID: ami-miR-215-5p
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-215
          Stem-loop Accession # MI0001203
          Stem-loop Sequence
          UCAGUAAGAACUGGUGUCCAGGAAAAUGACCUAUGAAUUGACAGACUGCUUUCAAAAUGUGCCUGUCAUUUCUAUAGGCCAAUAUUCUGUGCACUUUUCCUACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001134
          miRBase ID: gga-miR-215-5p
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-215
          Stem-loop Accession # MI0029538
          Stem-loop Sequence
          GUGUACAGGAAAAUGACCUAUGAAUUGACAGACUGUGUCUUCUAAAUUUGUCUGUCACUUCUGUAGGCCAAUAUUCUGCAUGCCUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037841
          miRBase ID: cpi-miR-215-5p
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-215
          Stem-loop Accession # MI0030690
          Stem-loop Sequence
          AUUCAGAAAUGGUGUACAGGAAAAUGACCUAUGAAUUGACAGACAAUGGGACUAAGUCUGUCUGUCAUUUCUGUAGGCCAAUAUUCUGUAUAUCUCUGCGACUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036060
          miRBase ID: chi-miR-215-5p
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-215
          Stem-loop Accession # MI0003031
          Stem-loop Sequence
          AUCAUUCAGAAAUGGUAUACGGGAAAAUGACCUAUGAAUUGACAGACAAUAUAGCUGAGUUUGUCUGUCAUUUCUUUAGGCCAAUAUUCUGUAUGACUGUGCUACUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002734
          miRBase ID: ggo-miR-215
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-215
          Stem-loop Accession # MI0012938
          Stem-loop Sequence
          AUCAUUAAGAAAUGGUAUACAGGAAAAUGACCUAUGAAUUGACAGACAAUAUGACUAAGUUUGUCUGUCAUUUCUGUAGGCCAAUAUUCUGUAUAUCUCUGCUACUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013190
          miRBase ID: eca-miR-215
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-215
          Stem-loop Accession # MI0003033
          Stem-loop Sequence
          AUCAUUAAGAAAUGGUAUACAGGAAAAUGACCUAUGAAUUGACAGACACUAUAGCUGAGUUUGUCUGUCAUUUCUUUAGGCCAAUAUUCUGUAUGACUGUGCUACUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002736
          miRBase ID: mne-miR-215
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-215
          Stem-loop Accession # MI0006783
          Stem-loop Sequence
          CAAUCCAGGAAGAAAUGGUGUACAGGAAAAUGACCUAUGAAUUGACAGACAGUAUAUUUAGAUUGUCUGUCAUUUCUGUAGGCCAAAGUUCUGCAUGCCUUCACUGCUCCGGCGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007000
          miRBase ID: oan-miR-215-5p
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-215
          Stem-loop Accession # MI0003027
          Stem-loop Sequence
          AUCAUUCAGAAAUGGUAUACGGGAAAAUGACCUAUGAAUUGACAGACAAUAUAGCUGAGUUUGUCUGUCAUUUCUUUAGGCCAAUAUUCUGUAUGACUGUGCUACUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002730
          miRBase ID: ptr-miR-215
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-215
          Stem-loop Accession # MI0010687
          Stem-loop Sequence
          AGAGAUGGUGUACAGGAAAAUGACCUAUGAAUUGACAGACAAUGUGGCUAAAUCUGUCUGUCAUUUCUGUAGGCCAAUAUUCUGUAUAUCUCUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010192
          miRBase ID: ssc-miR-215
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-215
          Stem-loop Accession # MI0003029
          Stem-loop Sequence
          AUCAUUCAGAAAUGGUAUACAGGAAAAUGACCUAUGAAUUGACAGACAAUACAGCUGAGUUUGUCUGUCAUUUCUUUAGGCCAAUAUUCUGUACAACUGUGCUACUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002732
          miRBase ID: ppy-miR-215
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-215
          Stem-loop Accession # MI0003025
          Stem-loop Sequence
          AUCAUUAAGAAAUGGUAUACAGGAAAAUGACCUAUGAAUUGACAGACACUAUAGCUGAGUUUGUCUGUCAUUUCUUUAGGCCAAUAUUCUGUAUGACUGUGCUACUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002728
          miRBase ID: mml-miR-215-5p
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-215
          Stem-loop Accession # MI0013804
          Stem-loop Sequence
          GUCCAGGAAAAUGACCUAUGAAUUGACAGACUGUCUUUUGCAUUUGCCUGUCACUUCUAUAGGCCAAUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014577
          miRBase ID: tgu-miR-215-5p
          Mature miRNA Sequence: AUGACCUAUGAAUUGACAGAC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03295053_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10874
          Ambion® Pre-miR™ miRNA Precursor : PM10874
          TaqMan™ MicroRNA Assay : 000518
          mirVana® miRNA mimic : MC10874
          TaqMan™ Advanced miRNA Assay : 478516_mir


          miRBase Alias:

          Human: hsa-miR-215 (v19)
          Gallus gallus: gga-miR-215 (v19)
          Macaca mulatta: mml-miR-215 (v19)
          Ochrobactrum anthropi: oan-miR-215 (v18)
          Trabulsiella guamensis ATCC 49490: tgu-miR-215 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline