Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM10978
          Assay Name: hsa-miR-301a-3p
          miRBase Accession Number: MI0000745
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 59151136 - 59151221 [-] on Build GRCh38
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Species: Human, Mouse, Rat, Anolis carolinensis, Callithrix jacchus, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Dog, Goat, Gorilla gorilla, Horse, Ictalurus punctatus, Microcebus murinus, Monodelphis domestica, Nomascus leucogenys, Ornithorhynchus anatinus, Otolemur garnettii, Pan troglodytes, Pongo pygmaeus, Python bivittatus, Rhesus monkey, Salmo salar, Taeniopygia guttata, Xenopus tropicalis
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM10978
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000034, mir-130

          Mature miRNA Sequence:

          CAGUGCAAUAGUAUUGUCAAAGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-301a-3p MIMAT0000688
          Mouse mmu-miR-301a-3p MIMAT0000379
          Rat rno-miR-301a-3p MIMAT0000552
          Anolis carolinensis aca-miR-301a-3p MIMAT0021916
          Callithrix jacchus cja-miR-301a MIMAT0049307
          Chrysemys picta cpi-miR-301a-3p MIMAT0037858
          View More Rows
          Cricetulus griseus cgr-miR-301a-3p MIMAT0023891
          Cyprinus carpio ccr-miR-301a MIMAT0026288
          Dog cfa-miR-301a MIMAT0009853
          Goat chi-miR-301a-3p MIMAT0036119
          Gorilla gorilla ggo-miR-301a MIMAT0049111
          Horse eca-miR-301a MIMAT0013032
          Ictalurus punctatus ipu-miR-301a MIMAT0029494
          Microcebus murinus mmr-miR-301a MIMAT0049153
          Monodelphis domestica mdo-miR-301-3p MIMAT0012739
          Nomascus leucogenys nle-miR-301a MIMAT0049397
          Ornithorhynchus anatinus oan-miR-301-3p MIMAT0006910
          Otolemur garnettii oga-miR-301a MIMAT0049593
          Pan troglodytes ptr-miR-301a MIMAT0008084
          Pongo pygmaeus ppy-miR-301a MIMAT0049335
          Python bivittatus pbv-miR-301a-3p MIMAT0039039
          Rhesus monkey mml-miR-301a-3p MIMAT0006257
          Salmo salar ssa-miR-301d-3p MIMAT0032599
          Taeniopygia guttata tgu-miR-301-3p MIMAT0014595
          Xenopus tropicalis xtr-miR-301 MIMAT0003635

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-301a
          Stem-loop Accession # MI0000745
          Stem-loop Sequence
          ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGG
          Chromosome Location Chr. 17 - 59151136 - 59151221 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000688
          miRBase ID: hsa-miR-301a-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: Chr. 17 - 59151136 - 59151221 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-301a
          Stem-loop Accession # MI0000401
          Stem-loop Sequence
          CCUGCUAACGGCUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCGAGCAGUGCAAUAGUAUUGUCAAAGCAUCCGCGAGCAGG
          Chromosome Location Chr. 11 - 87113004 - 87113089 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000379
          miRBase ID: mmu-miR-301a-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: Chr. 11 - 87113004 - 87113089 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-301a
          Stem-loop Accession # MI0000593
          Stem-loop Sequence
          CCUGCUGGCUACUGCUGACGACUGCUCUGACUUUAUUGCACUACUGUACUGUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCCGGGAGCAGGCUAC
          Chromosome Location Chr. 10 - 74417746 - 74417845 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000552
          miRBase ID: rno-miR-301a-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: Chr. 10 - 74417746 - 74417845 [+] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-301a
          Stem-loop Accession # MI0018844
          Stem-loop Sequence
          UUCCUGCUGCGGACAAGGGCUCUGACUUCAUUGCACUACUGUACUUCCCAACUAGCAGUGCAAUAGUAUUGUCAAAGCACUUGCAAGCAGAACUGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021916
          miRBase ID: aca-miR-301a-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-301a
          Stem-loop Accession # MI0040043
          Stem-loop Sequence
          UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049307
          miRBase ID: cja-miR-301a
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-301a
          Stem-loop Accession # MI0029550
          Stem-loop Sequence
          UGCUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037858
          miRBase ID: cpi-miR-301a-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-301a
          Stem-loop Accession # MI0020482
          Stem-loop Sequence
          GCUCGCGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCCGAAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023891
          miRBase ID: cgr-miR-301a-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-301a
          Stem-loop Accession # MI0023388
          Stem-loop Sequence
          ACAUCAAGUGCUGUUAACAGGUGCUCUGACUUCAUUGCACUACUGUAUUGGACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAGAGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026288
          miRBase ID: ccr-miR-301a
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-301a
          Stem-loop Accession # MI0010348
          Stem-loop Sequence
          CUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009853
          miRBase ID: cfa-miR-301a
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-301a
          Stem-loop Accession # MI0030729
          Stem-loop Sequence
          ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036119
          miRBase ID: chi-miR-301a-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-301a
          Stem-loop Accession # MI0039823
          Stem-loop Sequence
          UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049111
          miRBase ID: ggo-miR-301a
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-301a
          Stem-loop Accession # MI0012779
          Stem-loop Sequence
          ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013032
          miRBase ID: eca-miR-301a
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-301a
          Stem-loop Accession # MI0024611
          Stem-loop Sequence
          UCUGACUUCAUUGCACUACUGUAUUGGACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029494
          miRBase ID: ipu-miR-301a
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-301a
          Stem-loop Accession # MI0039868
          Stem-loop Sequence
          UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049153
          miRBase ID: mmr-miR-301a
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-301
          Stem-loop Accession # MI0012495
          Stem-loop Sequence
          ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012739
          miRBase ID: mdo-miR-301-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-301a
          Stem-loop Accession # MI0040137
          Stem-loop Sequence
          UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049397
          miRBase ID: nle-miR-301a
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-301
          Stem-loop Accession # MI0006727
          Stem-loop Sequence
          UAUUAUGGUGUGUUUUACGCUUUCUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUUCUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGAAUACACAGCAGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006910
          miRBase ID: oan-miR-301-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-301a
          Stem-loop Accession # MI0040374
          Stem-loop Sequence
          UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049593
          miRBase ID: oga-miR-301a
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-301a
          Stem-loop Accession # MI0008597
          Stem-loop Sequence
          ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008084
          miRBase ID: ptr-miR-301a
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-301a
          Stem-loop Accession # MI0040071
          Stem-loop Sequence
          UCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049335
          miRBase ID: ppy-miR-301a
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-301a
          Stem-loop Accession # MI0030300
          Stem-loop Sequence
          UUUUGCUGCUGACAAGUGCUCUGACUUCAUUGCACUACUGUACUUCCCAGCUAGCAGUGCAAUAGUAUUGUCAAAGCACUUGCGAGCAGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039039
          miRBase ID: pbv-miR-301a-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-301a
          Stem-loop Accession # MI0007685
          Stem-loop Sequence
          ACUGCUAACGAAUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAAAGCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006257
          miRBase ID: mml-miR-301a-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-301d
          Stem-loop Accession # MI0026673
          Stem-loop Sequence
          CUCUGACUUCAUUGCACUACUGUAUUGGACAGCUAGCAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032599
          miRBase ID: ssa-miR-301d-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-301
          Stem-loop Accession # MI0013823
          Stem-loop Sequence
          AGUGCUCUGACUUUAUUGCACUACUGUACUAUACAGAGAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGAGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014595
          miRBase ID: tgu-miR-301-3p
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-301-1
          Stem-loop Accession # MI0004876
          Stem-loop Sequence
          CUGCUAAUGUGUGCUCUGACUUUAUUGCACUACUGUACUAUACAGCUAGCAGUGCAAUAGUAUUGUCAAAGCAUCUGCAAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003635
          miRBase ID: xtr-miR-301
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA

          Stem-loop ID xtr-mir-301-2
          Stem-loop Accession # MI0004877
          Stem-loop Sequence
          GAAUUGCUCUGACAGUGUUGCACUACUGUGAUCUCAAAUGAAGCAGUGCAAUAGUAUUGUCAAAGCAUUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003635
          miRBase ID: xtr-miR-301
          Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306644_pri
          TaqMan™ Pri-miRNA Assay : Rn03464947_pri
          TaqMan™ Pri-miRNA Assay : Hs03303409_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10978
          TaqMan™ MicroRNA Assay : 000528
          mirVana® miRNA inhibitor : MH10978
          mirVana® miRNA mimic : MC10978
          TaqMan™ Advanced miRNA Assay : 477815_mir


          miRBase Alias:

          Human: hsa-miR-301 (v9.2)
          Human: hsa-miR-301a (v17)
          Mouse: mmu-miR-301a (v17)
          Mouse: mmu-miR-301 (v9.2)
          Rat: rno-miR-301a (v18)
          Aeromonas caviae Ae398: aca-miR-301a (v18)
          Macaca mulatta: mml-miR-301a (v19)
          Monodelphis domestica: mdo-miR-301 (v19)
          Ochrobactrum anthropi: oan-miR-301 (v18)
          Trabulsiella guamensis ATCC 49490: tgu-miR-301 (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline