Search
Search

| Assay Name: | hsa-miR-301a-3p |
|---|---|
| miRBase Accession Number: | MI0000745 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGUGCAAUAGUAUUGUCAAAGC |
| Species: | Human, Mouse, Rat, Anolis carolinensis, Callithrix jacchus, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Dog, Goat, Gorilla gorilla, Horse, Ictalurus punctatus, Microcebus murinus, Monodelphis domestica, Nomascus leucogenys, Ornithorhynchus anatinus, Otolemur garnettii, Pan troglodytes, Pongo pygmaeus, Python bivittatus, Rhesus monkey, Salmo salar, Taeniopygia guttata, Xenopus tropicalis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | CAGUGCAAUAGUAUUGUCAAAGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-301a-3p | MIMAT0000688 |
| Mouse | mmu-miR-301a-3p | MIMAT0000379 |
| Rat | rno-miR-301a-3p | MIMAT0000552 |
| Anolis carolinensis | aca-miR-301a-3p | MIMAT0021916 |
| Callithrix jacchus | cja-miR-301a | MIMAT0049307 |
| Chrysemys picta | cpi-miR-301a-3p | MIMAT0037858 |
| View More Rows | ||
| Cricetulus griseus | cgr-miR-301a-3p | MIMAT0023891 |
| Cyprinus carpio | ccr-miR-301a | MIMAT0026288 |
| Dog | cfa-miR-301a | MIMAT0009853 |
| Goat | chi-miR-301a-3p | MIMAT0036119 |
| Gorilla gorilla | ggo-miR-301a | MIMAT0049111 |
| Horse | eca-miR-301a | MIMAT0013032 |
| Ictalurus punctatus | ipu-miR-301a | MIMAT0029494 |
| Microcebus murinus | mmr-miR-301a | MIMAT0049153 |
| Monodelphis domestica | mdo-miR-301-3p | MIMAT0012739 |
| Nomascus leucogenys | nle-miR-301a | MIMAT0049397 |
| Ornithorhynchus anatinus | oan-miR-301-3p | MIMAT0006910 |
| Otolemur garnettii | oga-miR-301a | MIMAT0049593 |
| Pan troglodytes | ptr-miR-301a | MIMAT0008084 |
| Pongo pygmaeus | ppy-miR-301a | MIMAT0049335 |
| Python bivittatus | pbv-miR-301a-3p | MIMAT0039039 |
| Rhesus monkey | mml-miR-301a-3p | MIMAT0006257 |
| Salmo salar | ssa-miR-301d-3p | MIMAT0032599 |
| Taeniopygia guttata | tgu-miR-301-3p | MIMAT0014595 |
| Xenopus tropicalis | xtr-miR-301 | MIMAT0003635 |