Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11813
          Assay Name: hsa-miR-802
          miRBase Accession Number: MI0003906
          miRBase Version: v22.1
          Chromosome Location: Chr.21: 35720715 - 35720808 [+] on Build GRCh38
          Mature miRNA Sequence: CAGUAACAAAGAUUCAUCCUUGU
          Species: Human, Dog, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11813
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000353, mir-802

          Mature miRNA Sequence:

          CAGUAACAAAGAUUCAUCCUUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-802 MIMAT0004185
          Dog cfa-miR-802 MIMAT0009928
          Horse eca-miR-802 MIMAT0013183
          Pan troglodytes ptr-miR-802 MIMAT0008323
          Pig ssc-miR-802 MIMAT0041613
          Pongo pygmaeus ppy-miR-802 MIMAT0016110
          View More Rows
          Rhesus monkey mml-miR-802 MIMAT0006515

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-802
          Stem-loop Accession # MI0003906
          Stem-loop Sequence
          GUUCUGUUAUUUGCAGUCAGUAACAAAGAUUCAUCCUUGUGUCCAUCAUGCAACAAGGAGAAUCUUUGUCACUUAGUGUAAUUAAUAGCUGGAC
          Chromosome Location Chr. 21 - 35720715 - 35720808 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004185
          miRBase ID: hsa-miR-802
          Mature miRNA Sequence: CAGUAACAAAGAUUCAUCCUUGU
          Chromosome Location: Chr. 21 - 35720715 - 35720808 [+] on Build GRCh38

          Dog

          Stem-loop ID cfa-mir-802
          Stem-loop Accession # MI0010427
          Stem-loop Sequence
          UUCUAUUAUUUGUACUCAGUAACAAAGAUUCAUCCUUGUGUCCGUCAAGCAACAAGGAGAAUCUUUGUCACUUAGUGUAAUUAAUAGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009928
          miRBase ID: cfa-miR-802
          Mature miRNA Sequence: CAGUAACAAAGAUUCAUCCUUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-802
          Stem-loop Accession # MI0012928
          Stem-loop Sequence
          UUCUAUUAUUUGCAAUCAGUAACAAAGAUUCAUCCUUGUGUCCAUCAAGCAACAAGGAGAAUCUUUGUCACUUAGUGUAAUUAAUAGCUGCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013183
          miRBase ID: eca-miR-802
          Mature miRNA Sequence: CAGUAACAAAGAUUCAUCCUUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-802
          Stem-loop Accession # MI0008862
          Stem-loop Sequence
          UUCUGUUAUUUGCAGUCAGUAACAAAGAUUCAUCCUUGUGUCCAUCAUGCAACAAGGAGAAUCUUUGUCACUCAGUGUAAUUAAUAGCUGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008323
          miRBase ID: ptr-miR-802
          Mature miRNA Sequence: CAGUAACAAAGAUUCAUCCUUGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-802
          Stem-loop Accession # MI0033414
          Stem-loop Sequence
          CAGUAACAAAGAUUCAUCCUUGUGUCCACCAAGCAACAAGGAGGAUCUUUGUCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041613
          miRBase ID: ssc-miR-802
          Mature miRNA Sequence: CAGUAACAAAGAUUCAUCCUUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-802
          Stem-loop Accession # MI0015149
          Stem-loop Sequence
          GUUCUGUUAUUUGCAGUCAGUAACAAAGAUUCAUCCUUGUGUCCAUCACGCAGCAAGGAGAAUCUUUGUCACUUAGUGUAAUUAAUAGCUGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016110
          miRBase ID: ppy-miR-802
          Mature miRNA Sequence: CAGUAACAAAGAUUCAUCCUUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-802
          Stem-loop Accession # MI0007914
          Stem-loop Sequence
          GUUCUGUUAUUUGCAAUCAGUAACAAAGAUUCAUCCUUGUGUCCAUCAUGCAGCAAGGAGAAUCUUUGUCACUUAGUGUAAUUAAUAGCUGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006515
          miRBase ID: mml-miR-802
          Mature miRNA Sequence: CAGUAACAAAGAUUCAUCCUUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305020_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11813
          TaqMan™ MicroRNA Assay : 002004
          mirVana® miRNA inhibitor : MH11813
          mirVana® miRNA mimic : MC11813
          TaqMan™ Advanced miRNA Assay : 479181_mir

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline