Search
Search

| Assay Name: | hsa-miR-802 |
|---|---|
| miRBase Accession Number: | MI0003906 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGUAACAAAGAUUCAUCCUUGU |
| Species: | Human, Dog, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000353, mir-802 |
Mature miRNA Sequence: | CAGUAACAAAGAUUCAUCCUUGU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-802 | MIMAT0004185 | |
| Dog | cfa-miR-802 | MIMAT0009928 | |
| Horse | eca-miR-802 | MIMAT0013183 | |
| Pan troglodytes | ptr-miR-802 | MIMAT0008323 | |
| Pig | ssc-miR-802 | MIMAT0041613 | |
| Pongo pygmaeus | ppy-miR-802 | MIMAT0016110 | |
| View More Rows | |||
| Rhesus monkey | mml-miR-802 | MIMAT0006515 | |