Search
Search

| Assay Name: | hsa-miR-30b-3p |
|---|---|
| miRBase Accession Number: | MI0000441 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGGGAGGUGGAUGUUUACUUC |
| Species: | Human, Alligator mississippiensis, Chrysemys picta, Dasypus novemcinctus, Goat, Monodelphis domestica, Rhesus monkey |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | CUGGGAGGUGGAUGUUUACUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30b-3p | MIMAT0004589 |
| Alligator mississippiensis | ami-miR-30b-3p | MIMAT0038080 |
| Chrysemys picta | cpi-miR-30b-3p | MIMAT0037677 |
| Dasypus novemcinctus | dno-miR-30b-3p | MIMAT0047559 |
| Goat | chi-miR-30b-3p | MIMAT0036124 |
| Monodelphis domestica | mdo-miR-30b-3p | MIMAT0028551 |
| View More Rows | ||
| Rhesus monkey | mml-miR-30b-3p | MIMAT0026563 |