Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002129
          Assay Name: hsa-miR-30b*
          miRBase Accession Number: MI0000441
          miRBase Version: v22.1
          Chromosome Location: Chr.8: 134800520 - 134800607 [-] on Build GRCh38
          Mature miRNA Sequence: CUGGGAGGUGGAUGUUUACUUC
          Species: Human, Alligator mississippiensis, Chrysemys picta, Dasypus novemcinctus, Goat, Monodelphis domestica, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002129
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000005, mir-30

          Mature miRNA Sequence:

          CUGGGAGGUGGAUGUUUACUUC

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-30b-3p MIMAT0004589
          Alligator mississippiensis ami-miR-30b-3p MIMAT0038080
          Chrysemys picta cpi-miR-30b-3p MIMAT0037677
          Dasypus novemcinctus dno-miR-30b-3p MIMAT0047559
          Goat chi-miR-30b-3p MIMAT0036124
          Monodelphis domestica mdo-miR-30b-3p MIMAT0028551
          View More Rows
          Rhesus monkey mml-miR-30b-3p MIMAT0026563

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-30b
          Stem-loop Accession # MI0000441
          Stem-loop Sequence
          ACCAAGUUUCAGUUCAUGUAAACAUCCUACACUCAGCUGUAAUACAUGGAUUGGCUGGGAGGUGGAUGUUUACUUCAGCUGACUUGGA
          Chromosome Location Chr. 8 - 134800520 - 134800607 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004589
          miRBase ID: hsa-miR-30b-3p
          Mature miRNA Sequence: CUGGGAGGUGGAUGUUUACUUC
          Chromosome Location: Chr. 8 - 134800520 - 134800607 [-] on Build GRCh38

          Alligator mississippiensis

          Stem-loop ID ami-mir-30b
          Stem-loop Accession # MI0029691
          Stem-loop Sequence
          ACUGACUUUUAGUUCAUGUAAACAUCCUACACUCAGCUAUAACAACUGGAACGGCUGGGAGGUGGAUGUUUACUUCAACUGAUUUGAAAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038080
          miRBase ID: ami-miR-30b-3p
          Mature miRNA Sequence: CUGGGAGGUGGAUGUUUACUUC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-30b
          Stem-loop Accession # MI0029425
          Stem-loop Sequence
          UACUGAUUUUUAGUUCAUGUAAACAUCCUACACUCAGCUAUAAAACAUAGAACGGCUGGGAGGUGGAUGUUUACUUCAACUGACUUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037677
          miRBase ID: cpi-miR-30b-3p
          Mature miRNA Sequence: CUGGGAGGUGGAUGUUUACUUC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-30b
          Stem-loop Accession # MI0038935
          Stem-loop Sequence
          UGUAAACAUCCUACACUCAGCUGUGAUGUGUGGAUUGGCUGGGAGGUGGAUGUUUACUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047559
          miRBase ID: dno-miR-30b-3p
          Mature miRNA Sequence: CUGGGAGGUGGAUGUUUACUUC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-30b
          Stem-loop Accession # MI0030732
          Stem-loop Sequence
          ACCAAGUUUCCAGUUCAUGUAAACAUCCUACACUCAGCUGUAACACUUGAGUCGGCUGGGAGGUGGAUGUUUACUUCAGCUGACUUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036124
          miRBase ID: chi-miR-30b-3p
          Mature miRNA Sequence: CUGGGAGGUGGAUGUUUACUUC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-30b
          Stem-loop Accession # MI0023794
          Stem-loop Sequence
          UGUAAACAUCCUACACUCAGCUGUAACACAUGGAUCGGCUGGGAGGUGGAUGUUUACUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028551
          miRBase ID: mdo-miR-30b-3p
          Mature miRNA Sequence: CUGGGAGGUGGAUGUUUACUUC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-30b
          Stem-loop Accession # MI0002502
          Stem-loop Sequence
          ACCAAGUUUCAGUUCAUGUAAACAUCCUACACUCAGCUGUAAUACAUGGAUUGGCUGGGAGGUGGAUGUUUACUUCAGCUGACUUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026563
          miRBase ID: mml-miR-30b-3p
          Mature miRNA Sequence: CUGGGAGGUGGAUGUUUACUUC
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Human Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Human MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303066_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12436
          Ambion® Pre-miR™ miRNA Precursor : PM12436
          mirVana® miRNA inhibitor : MH12436
          mirVana® miRNA mimic : MC12436
          TaqMan™ Advanced miRNA Assay : 478804_mir
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          FGMIR : TaqManTM Human MicroRNA Array B v3
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0


          miRBase Alias:

          Human: hsa-miR-30b* (v17)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline