Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM19450
          Assay Name: hsa-miR-381-5p
          miRBase Accession Number: MI0000789
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101045920 - 101045994 [+] on Build GRCh38
          Mature miRNA Sequence: AGCGAGGUUGCCCUUUGUAUAU
          Species: Human, Rat
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM19450
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          AGCGAGGUUGCCCUUUGUAUAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-381-5p MIMAT0022862
          Rat rno-miR-381-5p MIMAT0017220

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-381
          Stem-loop Accession # MI0000789
          Stem-loop Sequence
          UACUUAAAGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGGAAUAUACAAGGGCAAGCUCUCUGUGAGUA
          Chromosome Location Chr. 14 - 101045920 - 101045994 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0022862
          miRBase ID: hsa-miR-381-5p
          Mature miRNA Sequence: AGCGAGGUUGCCCUUUGUAUAU
          Chromosome Location: Chr. 14 - 101045920 - 101045994 [+] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-381
          Stem-loop Accession # MI0003546
          Stem-loop Sequence
          AGCGAGGUUGCCCUUUGUAUAUUCGGUUUAUUGACAUGGGAUAUACAAGGGCAAGCUCUCU
          Chromosome Location Chr. 6 - 133876615 - 133876675 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017220
          miRBase ID: rno-miR-381-5p
          Mature miRNA Sequence: AGCGAGGUUGCCCUUUGUAUAU
          Chromosome Location: Chr. 6 - 133876615 - 133876675 [+] on Build Rnor_6.0


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464892_pri
          TaqMan™ Pri-miRNA Assay : Hs03303552_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19450
          TaqMan™ MicroRNA Assay : 463623_mat
          mirVana® miRNA inhibitor : MH19450
          mirVana® miRNA mimic : MC19450
          TaqMan™ Advanced miRNA Assay : rno478866_mir
          TaqMan™ Advanced miRNA Assay : 478866_mir


          miRBase Alias:

          Rat: rno-miR-381* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline