Search
Search

| Assay Name: | hsa-miR-381-5p |
|---|---|
| miRBase Accession Number: | MI0000789 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGCGAGGUUGCCCUUUGUAUAU |
| Species: | Human, Rat |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGCGAGGUUGCCCUUUGUAUAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-381-5p | MIMAT0022862 |
| Rat | rno-miR-381-5p | MIMAT0017220 |