Search
Search

| Assay Name: | mmu-miR-146b-3p |
|---|---|
| Stem-loop Accession Number: | MI0004665 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCCCUAGGGACUCAGUUCUGGU |
| Species: | Mouse, Cricetulus griseus |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000103, mir-146 |
Mature miRNA Sequence: | GCCCUAGGGACUCAGUUCUGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-146b-3p | MIMAT0004826 |
| Cricetulus griseus | cgr-miR-146b-3p | MIMAT0023776 |