Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481645_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-206-3p
          Stem-loop Accession Number: MI0000490
          miRBase Version: v22.1
          Chromosome Location: Chr.6: 52144349 - 52144434 [+] on Build GRCh38
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Astatotilapia burtoni, Bovine, Callithrix jacchus, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Dog, Gadus morhua, Gorilla gorilla, Guinea pig, Horse, Ictalurus punctatus, Macaca nemestrina, Metriaclima zebra, Neolamprologus brichardi, Ophiophagus hannah, Oreochromis niloticus, Ornithorhynchus anatinus, Pan troglodytes, Paralichthys olivaceus, Pongo pygmaeus, Pteropus alecto, Pundamilia nyererei, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Xenopus laevis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481645_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000038, mir-1

          Mature miRNA Sequence:

          UGGAAUGUAAGGAAGUGUGUGG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-206 MIMAT0000462
          Mouse mmu-miR-206-3p MIMAT0000239
          Rat rno-miR-206-3p MIMAT0000879
          Alligator mississippiensis ami-miR-206-3p MIMAT0038232
          Astatotilapia burtoni abu-miR-206 MIMAT0042094
          Bovine bta-miR-206 MIMAT0009260
          View More Rows
          Callithrix jacchus cja-miR-206 MIMAT0039491
          Chicken gga-miR-206 MIMAT0001139
          Chrysemys picta cpi-miR-206-3p MIMAT0037834
          Columba livia cli-miR-206-3p MIMAT0038589
          Cricetulus griseus cgr-miR-206 MIMAT0023846
          Cyprinus carpio ccr-miR-206 MIMAT0026264
          Dasypus novemcinctus dno-miR-206-3p MIMAT0047749
          Dog cfa-miR-206 MIMAT0006606
          Gadus morhua gmo-miR-206-3p MIMAT0044199
          Gorilla gorilla ggo-miR-206 MIMAT0024181
          Guinea pig cpo-miR-206-3p MIMAT0047104
          Horse eca-miR-206 MIMAT0013098
          Ictalurus punctatus ipu-miR-206 MIMAT0029461
          Macaca nemestrina mne-miR-206 MIMAT0002319
          Metriaclima zebra mze-miR-206 MIMAT0042332
          Neolamprologus brichardi nbr-miR-206 MIMAT0042508
          Ophiophagus hannah oha-miR-206 MIMAT0036819
          Oreochromis niloticus oni-miR-206 MIMAT0042696
          Ornithorhynchus anatinus oan-miR-206-3p MIMAT0006995
          Pan troglodytes ptr-miR-206 MIMAT0008066
          Paralichthys olivaceus pol-miR-206-3p MIMAT0025445
          Pongo pygmaeus ppy-miR-206 MIMAT0002318
          Pteropus alecto pal-miR-206-3p MIMAT0039798
          Pundamilia nyererei pny-miR-206 MIMAT0042897
          Python bivittatus pbv-miR-206-3p MIMAT0039011
          Rabbit ocu-miR-206-3p MIMAT0048328
          Rhesus monkey mml-miR-206 MIMAT0006237
          Salmo salar ssa-miR-206-3p MIMAT0032501
          Xenopus laevis xla-miR-206-3p MIMAT0046527
          Xenopus tropicalis xtr-miR-206 MIMAT0003623
          Zebrafish dre-miR-206-3p MIMAT0001866

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-206
          Stem-loop Accession # MI0000490
          Stem-loop Sequence
          UGCUUCCCGAGGCCACAUGCUUCUUUAUAUCCCCAUAUGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCGGCAAGUG
          Chromosome Location Chr. 6 - 52144349 - 52144434 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000462
          miRBase ID: hsa-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: Chr. 6 - 52144349 - 52144434 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-206
          Stem-loop Accession # MI0000249
          Stem-loop Sequence
          CCAGGCCACAUGCUUCUUUAUAUCCUCAUAGAUAUCUCAGCACUAUGGAAUGUAAGGAAGUGUGUGGUUUUGG
          Chromosome Location Chr. 1 - 20679010 - 20679082 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000239
          miRBase ID: mmu-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: Chr. 1 - 20679010 - 20679082 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-206
          Stem-loop Accession # MI0000948
          Stem-loop Sequence
          CUUCCCCAGGCCACAUGCUUCUUUAUAUCCUCAUAGAUAUCACUGCGCUAUGGAAUGUAAGGAAGUGUGUGGUUUUGGCAAGUG
          Chromosome Location Chr. 9 - 26791764 - 26791847 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000879
          miRBase ID: rno-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: Chr. 9 - 26791764 - 26791847 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-206
          Stem-loop Accession # MI0029793
          Stem-loop Sequence
          CUUCUCUUUUGAGGCAACAUGCUUCUUUAUAUCCCCAUAGGGAUAACAGUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCGGGGAGAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038232
          miRBase ID: ami-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Astatotilapia burtoni

          Stem-loop ID abu-mir-206
          Stem-loop Accession # MI0033791
          Stem-loop Sequence
          ACAUGCUUCCUUAUAUCCCCAUAUUAAUACGCCACUUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042094
          miRBase ID: abu-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-206
          Stem-loop Accession # MI0009772
          Stem-loop Sequence
          UGCUUCCCAAGGCCACAUGCUUCUUUAUAUCCCCAUACGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCGGCGAGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009260
          miRBase ID: bta-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-206
          Stem-loop Accession # MI0032016
          Stem-loop Sequence
          GGAUGACACUGCUUCCCGAGACCACAUGCUUCUUUAUAUCCCCAUAUGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCGGCAAGUGCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039491
          miRBase ID: cja-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-206
          Stem-loop Accession # MI0001207
          Stem-loop Sequence
          GAGAUGACAUGCUUCUUUAUAUCCCCAUAUGGAUUAGGCUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCAGGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001139
          miRBase ID: gga-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-206
          Stem-loop Accession # MI0029533
          Stem-loop Sequence
          UUCUCUUUUGAGACAACAUGCUUCUUUAUAUCCCCAUAUGAAUUAUGCUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCAGGGAGAUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037834
          miRBase ID: cpi-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-206
          Stem-loop Accession # MI0030013
          Stem-loop Sequence
          UUCUCUUAUGAGAUUACAUGCUUCUUUAUAUCCCCAUAGGGAUUAGGCUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCAGGGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038589
          miRBase ID: cli-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-206
          Stem-loop Accession # MI0020456
          Stem-loop Sequence
          UUCCCGAGGCCACAUGCUUCUUUAUAUCCGCAUAGAUAUCACUGCACUAUGGAAUGUAAGGAAGUGUGUGGUUUUGGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023846
          miRBase ID: cgr-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-206
          Stem-loop Accession # MI0023366
          Stem-loop Sequence
          UUGCUCUUUGAGAUGACAUACUUCCUUAUAUCCCCAUAUACGGUAAUAUCAAUCUAUGGAAUGUAAGGAAGUGUGUGGUCUCAAGNGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026264
          miRBase ID: ccr-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-206
          Stem-loop Accession # MI0039038
          Stem-loop Sequence
          ACAUGCUUCUUUAUAUCCCCAUACGGAUUACAUUGCUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047749
          miRBase ID: dno-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-206
          Stem-loop Accession # MI0008002
          Stem-loop Sequence
          ACAUGCUUCUUUAUAUCCCCAUACGGAUUACGUUGCUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006606
          miRBase ID: cfa-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-206
          Stem-loop Accession # MI0036116
          Stem-loop Sequence
          ACAUGCUUCCUUAUAUGCCCAUAUGAAUACGCCAGUUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044199
          miRBase ID: gmo-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-206
          Stem-loop Accession # MI0020728
          Stem-loop Sequence
          GCUGCUUCUGGAUGACACUGCUUCCCGAGGCCACAUGCUUCUUUAUAUCCCCAUAUGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCGGCAAGUGCCUCCUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024181
          miRBase ID: ggo-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-206
          Stem-loop Accession # MI0038699
          Stem-loop Sequence
          ACAUGCUUCUUUAUAUCCCCAUAGGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047104
          miRBase ID: cpo-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-206-2
          Stem-loop Accession # MI0012845
          Stem-loop Sequence
          AGGCCACAUGCUUCUUUAUAUCCCCAUACGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013098
          miRBase ID: eca-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-206-1
          Stem-loop Accession # MI0024564
          Stem-loop Sequence
          ACAUGCUUCCUUAUAUGCCCAUAUUAAUGCUCAAGUUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029461
          miRBase ID: ipu-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Stem-loop ID ipu-mir-206-2
          Stem-loop Accession # MI0024565
          Stem-loop Sequence
          ACAUACUUCCUUAUAUCCCCAUAUUCAGAAUUAAUCUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029461
          miRBase ID: ipu-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-206
          Stem-loop Accession # MI0002620
          Stem-loop Sequence
          UGCUUCCCGAGGCCACAUGCUUCUUUAUAUCCCCAUAUGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCGGCAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002319
          miRBase ID: mne-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-206
          Stem-loop Accession # MI0034101
          Stem-loop Sequence
          ACAUGCUUCCUUAUAUCCCCAUAUUAAUACGCCACUUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042332
          miRBase ID: mze-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-206
          Stem-loop Accession # MI0034344
          Stem-loop Sequence
          ACAUGCUUCCUUAUAUCCCCAUAUUAAUACGCCACUUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042508
          miRBase ID: nbr-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-206
          Stem-loop Accession # MI0031408
          Stem-loop Sequence
          UUUUGAGACAACACACUUCUUUAUAAUCCCAUAUGAAUUACAUUACUAUGGAAUGUAAGGAAGUGUGUGGUUUCAGAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036819
          miRBase ID: oha-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-206
          Stem-loop Accession # MI0034605
          Stem-loop Sequence
          ACAUGCUUCCUUAUAUCCCCAUAUUAAUACGCCACUUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042696
          miRBase ID: oni-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-206
          Stem-loop Accession # MI0006779
          Stem-loop Sequence
          AGGAGGCCGCCCCGGCCAACUGCUCUCCGAGACAACAUGCUUCUUUAUAUCCCCAUAUGGAUUACGCUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCGGGCCAGAGCCCUCCCGGCCGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006995
          miRBase ID: oan-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-206
          Stem-loop Accession # MI0008576
          Stem-loop Sequence
          GCUUCCCGAGGCCACAUGCUUCUUUAUAUCCCCAUAUGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCGGCAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008066
          miRBase ID: ptr-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Paralichthys olivaceus

          Stem-loop ID pol-mir-206
          Stem-loop Accession # MI0022206
          Stem-loop Sequence
          ACAUGCUUCCUUAUAUCCCCAUAUUCAUACAGCACUUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025445
          miRBase ID: pol-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-206
          Stem-loop Accession # MI0002619
          Stem-loop Sequence
          UGCUUCCCGAGGCCACAUGCUUCUUUAUAUCCCCAUAUGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCGGCAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002318
          miRBase ID: ppy-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-206
          Stem-loop Accession # MI0032334
          Stem-loop Sequence
          ACAUGCUUCUUUAUAUCCCCAUACGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039798
          miRBase ID: pal-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-206
          Stem-loop Accession # MI0034866
          Stem-loop Sequence
          ACAUGCUUCCUUAUAUCCCCAUAUUAAUACGCCACUUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042897
          miRBase ID: pny-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-206
          Stem-loop Accession # MI0030283
          Stem-loop Sequence
          UUUCUCUUUUUGAGGGGACACGCUUCUUUAUAAUCCCAUAUGAAUUACAUUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCAGAAAGAUGAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039011
          miRBase ID: pbv-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-206
          Stem-loop Accession # MI0039343
          Stem-loop Sequence
          ACAUGCUUCUUUAUAUCCCCAUACGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048328
          miRBase ID: ocu-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-206
          Stem-loop Accession # MI0007667
          Stem-loop Sequence
          UGCUUCCCGAGGCCACAUGCUUCUUUAUAUCCCCAUAUGGAUUACUUUGCUAUGGAAUGUAAGGAAGUGUGUGGUUUCGGCAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006237
          miRBase ID: mml-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-206
          Stem-loop Accession # MI0026600
          Stem-loop Sequence
          ACAUGCUUCCUUAUAUCCCCAUAUGAAUACUGCUGUUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032501
          miRBase ID: ssa-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-206
          Stem-loop Accession # MI0038292
          Stem-loop Sequence
          ACAUGCUUCUUUAUAUACCCAUAUGAACUACAUUGUUAUGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046527
          miRBase ID: xla-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-206
          Stem-loop Accession # MI0004863
          Stem-loop Sequence
          GAGGUCACAUGCUUCUUUAUAUACCCAUAUGAACUACAUUGUUAUGGAAUGUAAGGAAGUGUGUGGCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003623
          miRBase ID: xtr-miR-206
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-206-1
          Stem-loop Accession # MI0002045
          Stem-loop Sequence
          GAAUGUUGCCUCUUGUGAAGACAUGCUUCCUUAUAUGCCCAUAUUAAUGCUCAAGUUAUGGAAUGUAAGGAAGUGUGUGGUUUCAGGGGGAAAUUUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001866
          miRBase ID: dre-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA

          Stem-loop ID dre-mir-206-2
          Stem-loop Accession # MI0002046
          Stem-loop Sequence
          GAGUUGGCUGUUGCUUAUUGAGACGACAUACUUCCUUAUAUCCCCAUAUUCAGAAUUAAUCUAUGGAAUGUAAGGAAGUGUGUGGCUUCAGUGAGAUCUCAUCAUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001866
          miRBase ID: dre-miR-206-3p
          Mature miRNA Sequence: UGGAAUGUAAGGAAGUGUGUGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306546_pri
          TaqMan™ Pri-miRNA Assay : Hs03303328_pri
          TaqMan™ Pri-miRNA Assay : Rn03465364_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10409
          Ambion® Pre-miR™ miRNA Precursor : PM10409
          TaqMan™ MicroRNA Assay : 000510
          mirVana® miRNA inhibitor : MH10409
          mirVana® miRNA mimic : MC10409
          TaqMan™ Advanced miRNA Assay : 477968_mir
          TaqMan™ Advanced miRNA Assay : rno481438_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline