Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu482681_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-126a-3p
          Stem-loop Accession Number: MI0000471
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 136670602 - 136670686 [+] on Build GRCh38
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Callithrix jacchus, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Horse, Monodelphis domestica, Ornithorhynchus anatinus, Pan troglodytes, Pig, Pongo pygmaeus, Python bivittatus, Rhesus monkey, Taeniopygia guttata
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu482681_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000115, mir-126

          Mature miRNA Sequence:

          UCGUACCGUGAGUAAUAAUGCG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-126-3p MIMAT0000445
          Mouse mmu-miR-126a-3p MIMAT0000138
          Rat rno-miR-126a-3p MIMAT0000832
          Alligator mississippiensis ami-miR-126-3p MIMAT0038132
          Anolis carolinensis aca-miR-126-3p MIMAT0021731
          Callithrix jacchus cja-miR-126 MIMAT0039470
          View More Rows
          Chrysemys picta cpi-miR-126-3p MIMAT0037734
          Columba livia cli-miR-126-3p MIMAT0038500
          Cricetulus griseus cgr-miR-126a MIMAT0023745
          Dasypus novemcinctus dno-miR-126-3p MIMAT0047618
          Guinea pig cpo-miR-126-3p MIMAT0046971
          Horse eca-miR-126-3p MIMAT0013177
          Monodelphis domestica mdo-miR-126-3p MIMAT0026935
          Ornithorhynchus anatinus oan-miR-126-3p MIMAT0007126
          Pan troglodytes ptr-miR-126 MIMAT0007985
          Pig ssc-miR-126-3p MIMAT0018378
          Pongo pygmaeus ppy-miR-126 MIMAT0015750
          Python bivittatus pbv-miR-126-3p MIMAT0038919
          Rhesus monkey mml-miR-126 MIMAT0006183
          Taeniopygia guttata tgu-miR-126-3p MIMAT0014544

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-126
          Stem-loop Accession # MI0000471
          Stem-loop Sequence
          CGCUGGCGACGGGACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCCGUCCACGGCA
          Chromosome Location Chr. 9 - 136670602 - 136670686 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000445
          miRBase ID: hsa-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: Chr. 9 - 136670602 - 136670686 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-126a
          Stem-loop Accession # MI0000153
          Stem-loop Sequence
          UGACAGCACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCGGUCA
          Chromosome Location Chr. 2 - 26591357 - 26591429 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000138
          miRBase ID: mmu-miR-126a-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: Chr. 2 - 26591357 - 26591429 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-126a
          Stem-loop Accession # MI0000898
          Stem-loop Sequence
          UGACAGCACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGUGGUCA
          Chromosome Location Chr. 3 - 4042488 - 4042560 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000832
          miRBase ID: rno-miR-126a-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: Chr. 3 - 4042488 - 4042560 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-126
          Stem-loop Accession # MI0029724
          Stem-loop Sequence
          CGGCCCAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGCGGCCAGCACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038132
          miRBase ID: ami-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-126
          Stem-loop Accession # MI0018727
          Stem-loop Sequence
          CGCUCCCCGCGGUCCAUUAUUACUUUUGGUACGCGCUGUGCCACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGAGGGACAGCACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021731
          miRBase ID: aca-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-126
          Stem-loop Accession # MI0031994
          Stem-loop Sequence
          GCCUGGCCUCUGCUGGUGACAGGACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGUCCACAGCACCACAUCCAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039470
          miRBase ID: cja-miR-126
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-126
          Stem-loop Accession # MI0029462
          Stem-loop Sequence
          GGCGGCGACGCUGGUGCCGGCCCAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGCGGCCAGCCCUGGCGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037734
          miRBase ID: cpi-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-126
          Stem-loop Accession # MI0029958
          Stem-loop Sequence
          ACCGCUGGUGACGGCCCAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGUGGCCAGCACG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038500
          miRBase ID: cli-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-126a
          Stem-loop Accession # MI0020387
          Stem-loop Sequence
          CGGUGACAGCACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGUCAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023745
          miRBase ID: cgr-miR-126a
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-126
          Stem-loop Accession # MI0038968
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUGUGACGCUUCAAACUCGUACCGUGAGUAAUAAUGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047618
          miRBase ID: dno-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-126
          Stem-loop Accession # MI0038627
          Stem-loop Sequence
          CAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046971
          miRBase ID: cpo-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-126
          Stem-loop Accession # MI0012919
          Stem-loop Sequence
          CGACGGGACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCCGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013177
          miRBase ID: eca-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-126
          Stem-loop Accession # MI0012499
          Stem-loop Sequence
          GGGUCAUUAUUACUUUUGGUACGCGCUGUGACACAUCAAACUCGUACCGUGAGUAAUAAUGCGCCAUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026935
          miRBase ID: mdo-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-126
          Stem-loop Accession # MI0006861
          Stem-loop Sequence
          GAAGCAUCAGUGACAGCUGGCAACUGCCCAUUAUUACUUUUGGUACGCGCUGUGACGCUUCAAACUCGUACCGUGAGUAAUAAUGCGUGGCUGGCAGCACUGGCAGCGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007126
          miRBase ID: oan-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-126
          Stem-loop Accession # MI0008465
          Stem-loop Sequence
          GCUGGCGACGGGACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCCGUCCACGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007985
          miRBase ID: ptr-miR-126
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-126
          Stem-loop Accession # MI0016619
          Stem-loop Sequence
          CGACGGGACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018378
          miRBase ID: ssc-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-126
          Stem-loop Accession # MI0014818
          Stem-loop Sequence
          CGCUGGCGACGGGACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCCGUCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015750
          miRBase ID: ppy-miR-126
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-126
          Stem-loop Accession # MI0030218
          Stem-loop Sequence
          GCGGCUCAUUAUUACUUUUGGUACGCGCUGUGCCACUUCAAACUCGUACCGUGAGUAAUAAUGCGCGGAGGGCCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038919
          miRBase ID: pbv-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-126
          Stem-loop Accession # MI0007616
          Stem-loop Sequence
          CGCUGGUGAUGGGACAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGUCCACAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006183
          miRBase ID: mml-miR-126
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-126
          Stem-loop Accession # MI0013758
          Stem-loop Sequence
          AUCCUCGUUCCUGCAGGGCGGGUCUGGAGCCACGAGGAGCGGCGGGCGCCGCUGGUGACGGCCCAUUAUUACUUUUGGUACGCGCUGUGACACUUCAAACUCGUACCGUGAGUAAUAAUGCGCUGUGGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014544
          miRBase ID: tgu-miR-126-3p
          Mature miRNA Sequence: UCGUACCGUGAGUAAUAAUGCG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306244_pri
          TaqMan™ Pri-miRNA Assay : Hs03303230_pri
          TaqMan™ Pri-miRNA Assay : Rn03464585_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12841
          Ambion® Pre-miR™ miRNA Precursor : PM12841
          TaqMan™ MicroRNA Assay : 002228
          mirVana® miRNA inhibitor : MH12841
          mirVana® miRNA mimic : MC12841
          TaqMan™ Advanced miRNA Assay : 477887_mir
          TaqMan™ Advanced miRNA Assay : rno481318_mir


          miRBase Alias:

          Comamonas granuli NBRC 101663: cgr-miR-126 (v21)

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline