Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu482962_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-15a-5p
          Stem-loop Accession Number: MI0000069
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 50049119 - 50049201 [-] on Build GRCh38
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Species: Human, Mouse, Alligator mississippiensis, Ateles geoffroyi, Columba livia, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Horse, Lagothrix lagotricha, Lemur catta, Macaca nemestrina, Microcebus murinus, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Rabbit, Rhesus monkey, Saguinus labiatus, Xenopus laevis, Xenopus tropicalis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu482962_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000006, mir-15

          Mature miRNA Sequence:

          UAGCAGCACAUAAUGGUUUGUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-15a-5p MIMAT0000068
          Mouse mmu-miR-15a-5p MIMAT0000526
          Alligator mississippiensis ami-miR-15a-5p MIMAT0038036
          Ateles geoffroyi age-miR-15a MIMAT0002638
          Columba livia cli-miR-15a-5p MIMAT0038410
          Dasypus novemcinctus dno-miR-15a-5p MIMAT0047501
          View More Rows
          Gorilla gorilla ggo-miR-15a MIMAT0002640
          Guinea pig cpo-miR-15a-5p MIMAT0046849
          Horse eca-miR-15a MIMAT0013083
          Lagothrix lagotricha lla-miR-15a MIMAT0002656
          Lemur catta lca-miR-15a MIMAT0002648
          Macaca nemestrina mne-miR-15a MIMAT0002642
          Microcebus murinus mmr-miR-15a MIMAT0049138
          Pan paniscus ppa-miR-15a MIMAT0002646
          Pan troglodytes ptr-miR-15a MIMAT0002654
          Pongo pygmaeus ppy-miR-15a MIMAT0002652
          Rabbit ocu-miR-15a-5p MIMAT0048101
          Rhesus monkey mml-miR-15a-5p MIMAT0002650
          Saguinus labiatus sla-miR-15a MIMAT0002644
          Xenopus laevis xla-miR-15a-5p MIMAT0046470
          Xenopus tropicalis xtr-miR-15a MIMAT0003560

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-15a
          Stem-loop Accession # MI0000069
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location Chr. 13 - 50049119 - 50049201 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000068
          miRBase ID: hsa-miR-15a-5p
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: Chr. 13 - 50049119 - 50049201 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-15a
          Stem-loop Accession # MI0000564
          Stem-loop Sequence
          CCCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUGUUGAAAAGGUGCAGGCCAUACUGUGCUGCCUCAAAAUACAAGGA
          Chromosome Location Chr. 14 - 61632027 - 61632110 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000526
          miRBase ID: mmu-miR-15a-5p
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: Chr. 14 - 61632027 - 61632110 [-] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-15a
          Stem-loop Accession # MI0029666
          Stem-loop Sequence
          UACCUUGGCCUAACGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038036
          miRBase ID: ami-miR-15a-5p
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Ateles geoffroyi

          Stem-loop ID age-mir-15a
          Stem-loop Accession # MI0002945
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002638
          miRBase ID: age-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-15a
          Stem-loop Accession # MI0029906
          Stem-loop Sequence
          UUGGCAUAACGUAGCAGCACAUAAUGGUUUGUGGGUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038410
          miRBase ID: cli-miR-15a-5p
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-15a
          Stem-loop Accession # MI0038905
          Stem-loop Sequence
          UAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047501
          miRBase ID: dno-miR-15a-5p
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-15a
          Stem-loop Accession # MI0002947
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002640
          miRBase ID: ggo-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-15a
          Stem-loop Accession # MI0038561
          Stem-loop Sequence
          UAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046849
          miRBase ID: cpo-miR-15a-5p
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-15a
          Stem-loop Accession # MI0012829
          Stem-loop Sequence
          UAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013083
          miRBase ID: eca-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-15a
          Stem-loop Accession # MI0002963
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002656
          miRBase ID: lla-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Lemur catta

          Stem-loop ID lca-mir-15a
          Stem-loop Accession # MI0002955
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002648
          miRBase ID: lca-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-15a
          Stem-loop Accession # MI0002949
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002642
          miRBase ID: mne-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-15a
          Stem-loop Accession # MI0039851
          Stem-loop Sequence
          UAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049138
          miRBase ID: mmr-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-15a
          Stem-loop Accession # MI0002953
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002646
          miRBase ID: ppa-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-15a
          Stem-loop Accession # MI0002961
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002654
          miRBase ID: ptr-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-15a
          Stem-loop Accession # MI0002959
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002652
          miRBase ID: ppy-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-15a
          Stem-loop Accession # MI0039221
          Stem-loop Sequence
          UAGCAGCACAUAAUGGUUUGUGGAGUUUAAAAAGGUGCAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048101
          miRBase ID: ocu-miR-15a-5p
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-15a
          Stem-loop Accession # MI0002957
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002650
          miRBase ID: mml-miR-15a-5p
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-15a
          Stem-loop Accession # MI0002951
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002644
          miRBase ID: sla-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-15a
          Stem-loop Accession # MI0038261
          Stem-loop Sequence
          UAGCAGCACAUAAUGGUUUGUGGGUUACACAGAGGUGCAGGCCAUACUGUGCUGCCGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046470
          miRBase ID: xla-miR-15a-5p
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-15a
          Stem-loop Accession # MI0004799
          Stem-loop Sequence
          CCUUGACGUAAAGUAGCAGCACAUAAUGGUUUGUGGGUUACACAGAGGUGCAGGCCAUACUGUGCUGCCGCCAAAACACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003560
          miRBase ID: xtr-miR-15a
          Mature miRNA Sequence: UAGCAGCACAUAAUGGUUUGUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306795_pri
          TaqMan™ Pri-miRNA Assay : Hs03302582_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10235
          Ambion® Pre-miR™ miRNA Precursor : PM10235
          TaqMan™ MicroRNA Assay : 000389
          mirVana® miRNA inhibitor : MH10235
          mirVana® miRNA mimic : MC10235
          TaqMan™ Advanced miRNA Assay : 477858_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline