Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno480997_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-214-3p
          Stem-loop Accession Number: MI0000954
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 80130905 - 80131014 [+] on Build Rnor_6.0
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Species: Rat, Alligator mississippiensis, Ateles geoffroyi, Chicken, Fugu rubripes, Gadus morhua, Gorilla gorilla, Macaca nemestrina, Monodelphis domestica, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey, Saguinus labiatus, Tetraodon nigroviridis, Xenopus laevis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno480997_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000062, mir-214

          Mature miRNA Sequence:

          ACAGCAGGCACAGACAGGCAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-214-3p MIMAT0000885
          Alligator mississippiensis ami-miR-214-3p MIMAT0038239
          Ateles geoffroyi age-miR-214 MIMAT0002551
          Chicken gga-miR-214 MIMAT0007751
          Fugu rubripes fru-miR-214 MIMAT0003026
          Gadus morhua gmo-miR-214-3p MIMAT0044209
          View More Rows
          Gorilla gorilla ggo-miR-214 MIMAT0002550
          Macaca nemestrina mne-miR-214 MIMAT0002557
          Monodelphis domestica mdo-miR-214-3p MIMAT0004130
          Pan paniscus ppa-miR-214 MIMAT0002552
          Pan troglodytes ptr-miR-214 MIMAT0002554
          Pig ssc-miR-214-3p MIMAT0002147
          Pongo pygmaeus ppy-miR-214 MIMAT0002553
          Rhesus monkey mml-miR-214-3p MIMAT0002555
          Saguinus labiatus sla-miR-214 MIMAT0002556
          Tetraodon nigroviridis tni-miR-214 MIMAT0003027
          Xenopus laevis xla-miR-214-3p MIMAT0046535
          Xenopus tropicalis xtr-miR-214 MIMAT0003627
          Zebrafish dre-miR-214 MIMAT0001283

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-214
          Stem-loop Accession # MI0000954
          Stem-loop Sequence
          GUCCUGGAUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location Chr. 13 - 80130905 - 80131014 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000885
          miRBase ID: rno-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: Chr. 13 - 80130905 - 80131014 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-214
          Stem-loop Accession # MI0029797
          Stem-loop Sequence
          CGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCACUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038239
          miRBase ID: ami-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Ateles geoffroyi

          Stem-loop ID age-mir-214
          Stem-loop Accession # MI0002854
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002551
          miRBase ID: age-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-214
          Stem-loop Accession # MI0008208
          Stem-loop Sequence
          GGACUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007751
          miRBase ID: gga-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-214
          Stem-loop Accession # MI0003356
          Stem-loop Sequence
          CAGGAUGAGACUGUUGUGGUGUGUCUGCCUGUCUACACUUGCUGUGCAGACCUUCUGCUCCUGUACAGCAGGCACAGACAGGCAGACAGAUGGCAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003026
          miRBase ID: fru-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-214-1
          Stem-loop Accession # MI0036126
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGACAUUCUGCUCCUGUACAGCAGGCACAGACAGGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044209
          miRBase ID: gmo-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Stem-loop ID gmo-mir-214-2
          Stem-loop Accession # MI0037985
          Stem-loop Sequence
          UGCCUGUCUUCACUUGCUGUGCAGUAAUUGGUUCAGCCUGUACAGCAGGCACAGACAGGCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044209
          miRBase ID: gmo-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-214
          Stem-loop Accession # MI0002853
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002550
          miRBase ID: ggo-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-214
          Stem-loop Accession # MI0002860
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002557
          miRBase ID: mne-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-214
          Stem-loop Accession # MI0005319
          Stem-loop Sequence
          GGGCUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004130
          miRBase ID: mdo-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-214
          Stem-loop Accession # MI0002855
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002552
          miRBase ID: ppa-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-214
          Stem-loop Accession # MI0002857
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002554
          miRBase ID: ptr-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-214
          Stem-loop Accession # MI0002441
          Stem-loop Sequence
          GGCCUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002147
          miRBase ID: ssc-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-214
          Stem-loop Accession # MI0002856
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002553
          miRBase ID: ppy-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-214
          Stem-loop Accession # MI0002858
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002555
          miRBase ID: mml-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-214
          Stem-loop Accession # MI0002859
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002556
          miRBase ID: sla-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-214
          Stem-loop Accession # MI0003357
          Stem-loop Sequence
          GUGGUGUGUCUGCCUGUCUACACUUGCUGUGCAGACCUUCUGCUCCUGUACAGCAGGCACAGACAGGCAGACAGAUGGCAGCCCCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003027
          miRBase ID: tni-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-214-1
          Stem-loop Accession # MI0038296
          Stem-loop Sequence
          GCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046535
          miRBase ID: xla-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Stem-loop ID xla-mir-214-2
          Stem-loop Accession # MI0038297
          Stem-loop Sequence
          GCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046535
          miRBase ID: xla-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-214
          Stem-loop Accession # MI0004867
          Stem-loop Sequence
          UUGGCUGGAAGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003627
          miRBase ID: xtr-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-214
          Stem-loop Accession # MI0001381
          Stem-loop Sequence
          UGACUGAGAGCGUUGUCUGUCUGCCUGUCUACACUUGCUGUGCAGAACUUCCUGCACCUGUACAGCAGGCACAGACAGGCAGACAGAUGGCAGCCCGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001283
          miRBase ID: dre-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465830_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10347
          Ambion® Pre-miR™ miRNA Precursor : PM10347
          TaqMan™ MicroRNA Assay : 000517
          mirVana® miRNA inhibitor : MH10347
          mirVana® miRNA mimic : MC10347


          miRBase Alias:

          Staphylococcus schleiferi: ssc-miR-214 (v21)

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline