Search
Search

| Assay Name: | rno-miR-344g |
|---|---|
| Stem-loop Accession Number: | MI0021837 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGUCAGGCUCCUGGCAGGAGUC |
| Species: | Rat |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000267, mir-344 |
Mature miRNA Sequence: | AGUCAGGCUCCUGGCAGGAGUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-344g | MIMAT0025052 |