Search
Search

| Assay Name: | dme-miR-276* |
|---|---|
| miRBase Accession Number: | MI0000359 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGCGAGGUAUAGAGUUCCUACG |
| Species: | Drosophila melanogaster, Drosophila grimshawi, Drosophila willistoni |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000124, mir-276 |
Mature miRNA Sequence: | CAGCGAGGUAUAGAGUUCCUACG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-276a-5p | MIMAT0000336 |
| Drosophila melanogaster | dme-miR-276b-5p | MIMAT0020815 |
| Drosophila grimshawi | dgr-miR-276a-5p | MIMAT0008575 |
| Drosophila willistoni | dwi-miR-276a-5p | MIMAT0009007 |